Lab Equipment Demo: Introducing the Cole-Parmer 8000M and 8000D Mixer/Mill® High-Energy Ball Mills
Presenting theIntroducing the Cole-Parmer 8000M and 8000D Mixer/Mill® High-Energy Ball Mills
Description
See how these 8000 Series Mixer/Mills, high-energy ball mills, pulverize rocks, sand, minerals, slag, ceramics, and other brittle to hard samples from 0.5 g to 25 g, reducing them to analytical fineness. Learn more about the 8000M Mixer/Mills single chamber and the 8000D dual-chamber Mixer/Mills. You won't believe how many times these Mixer/Mills can shake per minute.