Wipes Selection Guide
Keep your areas clean and reduce the spread of germs by simply grabbing a cleaning wipe. We offer a variety of dry and presaturated wipes for standard labs as well as clean rooms.
Cleanroom wipes decrease the risk of contamination in controlled environments. These specialized wipes have features such as individually sealed packaging, easy dispensing canisters, sealed edges to eliminate fiber release, durability, maximum performance, excellent pickup and retention of particles, and gamma irradiated to a 10 to the negative six sterility assurance level. We even have wipes in compliance with ISO Class 3, 5, 6 and 8.
To help you select the correct wipe for your lab, we have compiled the features of various wipe types as well as their performance for specific application areas (see Table 1).
Table 1. Wipes Selection Guide
| Type | Format | Features | GSM | Applications |
| Support Areas | Gowning | General Wiping Primary Areas | Work Stations / Equipment | Aseptic Core, Fill and Sterile Areas |
| Quilted two-ply polyester | Nonsterile | Highly absorbent Ultra clean Laser-sealed edges Cleanroom laundered | 250 ±5 g/m2 | ✓ | ✓ | ✓ | ✓✓ | |
| Sterile | Highly absorbent Ultra clean Laser-sealed edges Cleanroom laundered Gamma sterile | 250 ±5 g/m2 | ✓ | ✓ | ✓ | ✓✓ | ✓✓ |
| Single-ply polyester | Nonsterile | Good chemical resistance Low NVRS, fibers, ions, and extractables Cleanroom laundered | 135 ±5 g/m2 | ✓ | ✓ | ✓ | ✓✓ | |
| Sterile | Good chemical resistance Low NVRS, fibers, ions and extractables Cleanroom Laundered Gamma sterile | 135 ±5 g/m2 | ✓ | ✓ | ✓ | ✓✓ | ✓✓ |
| Microfiber | Nonsterile | Excellent residue removal Ideal for glass, optics, or similar | 100 ±5 g/m2 | ✓ | ✓ | ✓ | ✓✓ | |
| Polycellulose | Nonsterile | Excellent absorbency Polyester gives needed cleanliness levels Excellent for spills | 68 ±5 g/m2 | ✓ | ✓ | ✓ | ✓ | |
| Sterile | Excellent absorbency Polyester gives needed cleanliness levels Excellent for spills Gamma sterile | 68 ±5 g/m2 | ✓ | ✓ | ✓ | ✓ | |
| Polypropylene, wood pulp, viscose blend; presaturated | Nonsterile | Premoistened with 70% alcohol or 100% DI water Strong, nonwoven cloth, soft, and lint-free Excellent for cleaning surfaces/removing residue | 40 g/m2 | ✓ | ✓ | ✓ | ✓ | |
| Presaturated polyester | Sterile | Premoistened with 70% IPA and 30% DI/WFI 2 120 GSM, laser-cut edge reduces fiber contamination Very low levels of particulate and extractable counts High-abrasion resistance Gamma sterile | 120 ±5 g/m2 | ✓ | ✓ | ✓ | ✓✓ | ✓✓ |
| Nonsterile | Premoistened with 70% IPA and 30% DI Ideal for quick removal of surface-level contaminants from workbenches, electronics, equipment and tools in ISO Class 8 | 45 ±5 g/m2 | ✓ | ✓ | ✓ | ✓ | |
| Presaturated polycellulose | Sterile | 55% cellulose, 45% polyester blend Premoistened with 70% isopropyl alcohol and 30% DI/WFI Nonwoven, with no chemical binders Cold knife cut edge Gamma sterilized and packaged in an ISO Class 6 cleanroom Convenient resealable pouch | 56 ±5 g/m2 | ✓ | ✓ | ✓ | ✓ | |
| | | | ✓ = suitable ✓✓=preferred |