Chromatography Column and Phase Cross-Reference
Phase Desription Similar Phase Similar column
Non-polar
Dimethylpolysiloxane OV-1, OV-101, SE-30
DB-1 SPB-1 HP-1 RTx-1 CPSil 5CB AT-1 BP-1
(5% diphenyl)
dimethylpolysiloxane
SE-52, SE-54, OV-73
DB-5 SPB-5 HP-5 RTx-5 CPSil 8CB AT-5 BP-5
Midpolarity
(14% cyanopropylphenyl)
methylpolysiloxane
OV-1701
DB-1701 SPB-1701 RTx-1701 CPSil 19CB AT-1701 BP-10
(50% phenyl)
methylpolysiloxane
OV-17
DB-17 SPB-2250 HP-50 RTx-50 AT-1701
Polar
(50% cyanopropylphenyl)
methylpolysiloxane
OV-225
DB-225 SPB-2330 HP-225 RTx-225 CPSil 43CB AT-225 BP-225
Polyethylene glycol(PEG) Carbowax 20 M
DB-WAX SUPEL
COWAX 10
HP-(iNNO)Wax Stabil
wax
CP-Wax 52CB AT-WAX BP-20
(78% cyanopropyl)
methylpolysiloxane
Silar 7CP,SP-2310
DB-23 SP-2230 RTx-2330 CPSil 88CB AT-Silar BPX-70
Learn more about Chromatography: What is Chromatography and How it Does it Work?