Slimmer. Simpler. Faster
Oakton® PH550 pH Meter & CON550 Conductivity Meter
Cole-Parmer, Vernon Hills, Ill—The Oakton family has some new additions of compactly designed and economical meters for benchtop pH and conductivity measurement. The meters have IP54-rated housing which is dust- and spill-proof, while connectors are protected by silicone seal caps. Both meters have the capacity to automatically recognize NIST and USA calibration solutions and can store and recall up to 50 groups of data. What’s more, these meters have a warranty of up to 3 years.
The PH 550 benchtop pH meter features a microprocessor chip which provides automatic temperature compensation, data storage, parameter setup, and Max/Min reading display. The meter offers a pH range from 0 to 14 and a three-point automatic calibration. The advanced digital processing technology improves meter response time and measuring accuracy. A stable reading display icon appears when the measuring value is stable. The 3-in-1 combination pH electrode measures pH and temperature simultaneously. The compact meter includes a detachable electrode holder and three bottles of pH buffer solution.
The CON 550 benchtop conductivity meter features a microprocessor chip which provides automatic calibration, automatic temperature compensation, data storage, parameter setup, and Max/Min reading display. The meter reads 0 to 200 mS/cm across four ranges and the four-point conductivity calibration. The advanced digital processing technology improves meter response time and measuring accuracy. A stable reading display icon appears when the measuring value is stable. A conductivity electrode with temperature sensor measures conductivity and temperature simultaneously and allows for automatic temperature compensation. The compact meter includes a detachable electrode holder and three bottles of conductivity calibration solution.