Environmental Express Oakton CON 550 Benchtop Conductivity Meter Kit

Representative image only.

Environmental Express Oakton CON 550 Benchtop Conductivity Meter Kit

Economical meter for benchtop conductivity measurement

₹83,600.00INR / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹83,600.00INR / Each

Buy It Together

Current Item

Product Description

Includes

Plastic combination conductivity electrode (K=1.0) with temperature sensor, electrode holder, conductivity standard calibration solutions (84 µS/1413 µS/12.88 mS, 50 mL each), and 9 VAC power adapter.

Key Features

  • Automatically recognizes calibration solutions
  • Compact design saves valuable benchtop space
  • Data logging allows you to store and recall up to 50 groups of data
  • IP54-rated housing is dust- and spill-proof
  • 3 year warranty

More About this Item

The CON 550 benchtop conductivity meter features a microprocessor chip which provides automatic calibration, automatic temperature compensation, data storage, parameter setup, and Max/Min reading display. The meter reads 0 to 200 mS/cm across four ranges and the four-point conductivity calibration, with automatic solution recognition, makes for easy calibration. The advanced digital processing technology improves meter response time and measuring accuracy. A stable reading display icon appears when the measuring value is stable.

A conductivity electrode with temperature sensor measures conductivity and temperature simultaneously and allows for automatic temperature compensation. The compact meter includes a detachable electrode holder and three bottles of conductivity calibration solution. It has a dustproof and spillproof rating of IP54. Connectors are protected by silicone seal caps.

3 Year WarrantyConformite EuropeenneIP54 Rated
Specifications
  • Meter TypeBenchtop
  • Display typeLCD
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 4 points
  • Min Conductivity0 µS/cm
  • Max Conductivity200 mS/cm
  • Conductivity Resolution0.1/1 µS/cm; 0.01/0.1 mS/cm
  • Conductivity Accuracy±1.0% full-scale ±1 digit
  • Min TDS0
  • Max TDS100 g/L
  • TDS Resolution0.1/1 mg/L, 0.01/0.1 gL
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Temperature Resolution0.1/1°F (0.1°C)
  • Temperature Accuracy±0.5°C ±1 digit
  • Temperature compensation Automatic or manual from 32 to 122°F (0 to 50°C)
  • Data Logging (points)50 groups of data
  • Length (cm)23.32
  • Width (cm)15.24
  • Height (cm)6.35
  • Width (in)6
  • Length (in)8
  • Height (in)2 1/2
  • DescriptionBenchtop Conductivity Meter Kit with Probe and Stand
  • Warranty3 year
Reviews
Q&A