New Environmental Express® AutoBlock® Fill Automates Dangerous Process in Metals Digestion
September 14, 2017, Charleston, SC—The latest generation of the AutoBlock, from Environmental Express, safely and accurately automates the most dangerous step of sample prep for metals digestion—the addition of acids and reagents. The accessory fits any size HotBlock® digestion system to provide automation at an affordable price. The system is ideal for any laboratory performing their own unique metals digestion method.
The AutoBlock Fill dispenses from up to five reagent bottles at a time. A five-way valve eliminates the need to swap out reagent bottles when switching digestion methods, providing easy switching between reagents. Included software directs the AutoBlock Fill to dispense reagent as programmed by user. Reagent is dispensed via an integrated Masterflex® peristaltic pump accurate to ±2% volume and can be calibrated to any dispensing volume. The AutoBlock Fill can be controlled via the onboard touch screen or by directly connecting a computer with the included software
The metals digestion process is extremely corrosive and the AutoBlock Fill unit fits inside of standard-sized fume hoods or the Environmental Express AirLite™ Benchtop Enclosure to provide the safest and cleanest environment to handle acidic fumes. The AutoBlock Fill is constructed from the same acid-resistant materials as the company’s HotBlock systems.
For more information on the AutoBlock Fill, please visit EnvExp.com or call 1-800-343-5319.
Environmental Express, a Cole-Parmer company, is a leading developer, manufacturer and distributor of environmental laboratory equipment and consumable supplies for commercial, governmental, industrial and academic laboratories worldwide. The company provides an entire range of laboratory products used in applications such as water/wastewater analysis, oil and grease analysis, metals analysis and hazardous waste analysis. We pride ourselves on providing innovative products, superior technical support, knowledgeable customer service and expedited shipping.
Popular Categories
- Balances and Scales
- Analytical Balances
- Bench and Floor Scale Bases
- Bench and Floor Scales
- Counting Scales
- Crane and Hanging Scales
- Health Scales
- Indicators for Scales
- Mechanical Balances
- Moisture Balances
- Platform and Pallet Scales
- Production Weighing
- Semi and Micro Analytical Balances
- Toploading Balances
- Weighing Accessories
- Weighing Dishes
- Calibration
- Electrical Products
- Batteries and Battery Testers
- Cable Testers
- Cable Ties
- Clamp Meters
- Electrical Accessories
- Electronics Enclosures
- Function Generators
- Gauss Meters
- Insulation Testers
- LCR Meters
- Megohmmeters
- Motor Rotation Testers
- Multimeters
- Oscilloscopes
- Outlet Strips
- Power Meters and Analyzers
- Power Supplies
- Resistance Testers
- RF Signal Generators
- Static Control Products
- Surge Protectors
- Test Leads and Probes
- Voltage Detectors
- Filtration Products
- Fluid Handling
- Furniture
- Heating and Cooling Equipment
- Laboratory Equipment
- Baths and Circulators
- Burettes (Burets)
- Calorimeters
- Centrifuges
- Chromatography Products
- Desiccators
- Digestion Systems
- Dispensers
- Distillation
- Electrophoresis Products
- Electroporation Products
- Environmental Samplers
- Extraction Apparatus
- Fermentation Products
- Flame Photometers
- Fluid Bed Dryers
- Freeze Dryers
- Glassware Washers
- Histology Products
- Homogenizers and Ultrasonic Processors
- Humidifiers and Dehumidifiers
- Lab Shakers
- Laboratory Evaporators
- Laboratory Sieves and Sieve Shakers
- Leachate Equipment
- Lighting and Illumination
- Magnetic Stirrers and Hot Plates
- Microscopes and Magnification
- Mixers and Blenders
- Particle Counters
- Pipettes and Pipettors
- Refractometers
- Thermal Cyclers
- Titrators
- Ultrasonic Cleaners
- Vacuums
- Water Purification Products
- Laboratory Supplies
- Air Sampling
- Animal Research Products
- Bags
- Beakers
- Books and Reference Materials
- Bottles and Jars
- Calculators
- Carboys
- Chemistry Kits
- Clean Room Stationery
- Containers
- Cuvettes
- Dialysis Products
- Dippers
- Distillation Supplies
- Field Sampling
- Flasks
- Graduated Cylinders
- Ice Buckets and Ice Pans
- Lab Cleaning Products
- Lab Organizers
- Labels and Label Printers
- Laboratory Dishes
- Laboratory Timers and Clocks
- Labware
- Microbiology Products
- Microplate Products
- Mortars and Pestles
- Parts and Accessories
- Racks
- Sampling Wipes
- Shippers
- Syringes
- Tubes
- Vials
- Watch Glasses
- Material Handling and Storage
- Material Testing and Physical Testing
- Pressure and Vacuum Instruments
- Process Controllers
- Pumps
- Safety Products
- Air Quality Monitors
- Arc Flash Products
- Combustion Analyzers
- Eyewash
- Eyewear and Safety Glasses
- Floor Mats and Matting
- Gas Detectors
- Gloves
- Hearing Protection
- Lockout and Tagout Product
- Personal Monitor Badges
- Radiation Detectors
- Refrigerant Products
- Respirators and Masks
- Safety Apparel
- Safety Shields
- Safety Signs and Labels
- Solvent Safety
- Sound Meters
- Spill Control
- Temperature Measurement Products
- Test and Measurement Products
- Water Quality Products
- Colorimeters
- Colorimetry Reagents
- Conductivity, Resistivity, and TDS Meters
- Dissolved Oxygen Meters
- Dye Tracers
- Electrodes
- Flocculation Testers
- Multiparameter Meters
- pH Buffers
- pH Meters
- Turbidity Accessories
- Turbidity Meters
- Water Quality Accessories
- Water Quality Standards, and Solutions
- Water Quality Test Strips