Masterflex® Peristaltic Tubing Pumps
Masterflex® is now part of Avantor® CONTINUE TO MASTERFLEX.COM >
Engineered for long life, Masterflex® peristaltic pump systems provide superior accuracy and repeatability and ensure optimal performance when used with the proper tubing. These pumps offer the latest technology including MasterflexLive® cloud-enabled connectivity, easy control and tubing changes, and the low-maintenance and cost-effectiveness of peristaltic pumps.
To provide you with an easier way to select the fine pumps we offer, Ismatec pumps are now a part of the Masterflex family of products! The Ismatec Series pumps fit into the Masterflex brand offering as the microflow, high accuracy peristaltic pumps. Choose from a wide range of flows with the Masterflex Ismatec Series, Masterflex L/S Series, Masterflex I/P Series and Masterflex B/T Series peristaltic pumps. You can also find many tubing formulations compatible with the pumps we offer. Keep in mind that Ismatec pumps accept Ismatec pump tubing and Masterflex pumps accept Masterflex pump tubing for clean fluid paths. With over 10,000 pump options and many tubing formulations, you can find what you need from the Masterflex brand.
LIVE webinar - register today
Top Reasons to choose Masterflex
  • 1EASY
    • Quick installation.
    • Low maintenance.
    • Easy tubing changeover.
  • 2 Durable
    • Trusted for over 55 years.
    • Vast chemical compatibility.
  • 3 Versatile
    • From research to process, handles almost any fluid.
    • Multichannel capabilities.
  • 4 Customizable
    • Configurable systems
    • Many tubing options.
    • Easy scale-up.
  • 5 Accurate
    • Flow ranges - 0.0001 mL/min to 42 L/min.
    • Rigorous design standards.
Masterflex peristaltic pump series — pick your pump
Ismatec high accuracy microflow peristaltic pumps
Ismatec® Series
Microflow, high accuracy
Flow Range:
0.0001 to 365 mL/min
Masterflex L/S precise flow peristaltic pump
L/s® Series
High accuracy, precise flow
Flow Range:
0.00005 to 3400 mL/min
Masterflex I/P powerfil peristaltic pump
i/p® Series
Powerful systems
Flow Range:
0.001 to 26 LPM
Masterflex B/T highest flow rates peristaltic pumps
b/t® Series
Highest flow rates
Flow Range:
0.3 to 42 LPM
NEW Masterflex Ultrasonic Flow Sensor
Masterflex
Shop the best selection of Peristaltic Pumps available.
Microflow
high-accuracy
powerful systems
highest flow rate
oem design
Custom product development
Expert peristaltic pump system development.
Remote Pump Monitoring
MasterflexLIVE and Masterflex peristaltic pumps
24/7 Real-time monitoring
Choose the Right Masterflex Tubing Formulation
find your perfect tubing formulation
Chemical Compatibility Search
chemical compatibility
Related Services
Calibration Services
Innocal
Save time & money using our in-house Instrument Calibration Specialists. NIST-traceable calibration.
CUSTOM CUT TUBING
CUSTOM CUT-to-length TUBING
We have the flexibility to meet your needs AND save you time & money! Choose type, length & quantity.
Masterflex Single-Use Sterile Bioprocess Assemblies
Masterflex Single-Use
Your source for Disposable Sterile Bioprocess Assemblies
Need Help with this product?
Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.
Call Us At91-22-61394444
Live chat