The Kinesis TELOS MicroPlate – Small but significant, simple yet sophisticated
October 2018 — In today’s bioanalytical laboratory, managing waste is a hot topic. We, as a human race, have to rationalise which consumables we use, whilst weighing up their practicality in everyday use. At Cole-Parmer we strive to consciously design our products, where possible, with both you and the environment in mind.
Working with small sample volumes of biological fluids requires an optimal and cost-effective solution. TELOS MicroPlate, which is comprised of a base-plate populated with individual wells, is a fine example of where design has successfully met the requirements of versatility, cost-effectiveness, high throughput and a reduction in sample loss. This is achieved through several key features including well design, collection plate and vacuum manifold compatibility, and modular flexibility.
Each well is consciously designed with a tapered outlet containing a sorbent and frit bed (10 - 15μl). This maximises analyte and sorbent interactions, keeping elution volumes to a minimum, which is particularly important for small sample volumes (≥100μl), ensuring minimal evaporation times prior to analysis by LC-MS/MS. The wells are populated into the base-plate, with an audibly secure ‘click’, of the TELOS MicroPlate, however, the amount used can be flexibly altered and varied according to the protocol being undertaken, whether this is a high throughput assay or for method development. Unrequired ports on the base-plate are isolated using sealing strips. Once the wells are in place, the outlets from the TELOS MicroPlate are designed in such a way as to give good collection plate penetration, preventing well to well cross-contamination.
Overall the format that the TELOS MicroPlate allows you to optimise your plate by matching the sample numbers to be processed. Its design minimises evaporation times, prevents cross-contamination, whilst improving throughput. It is made of small components designed with simplicity in mind, which fit together to create a sophisticated, and yet a cost-effective piece of technology that can significantly impact your protocols on a daily basis
For more information, you can request a brochure and a sorbent selection poster from [email protected] or contact Matthew Cleeve, TELOS Category Manager, [email protected].
W: Coleparmer.com
E: [email protected]
T: +44(0)1480 272279
About Cole-Parmer
Cole-Parmer is a leading global source of test & measurement, biosciences and fluid handling products, instrumentation, equipment and supplies. We are proven experts in the fields of temperature measurement and control, electrochemistry and chromatography. Our portfolio of unique industry-leading brands includes Argos Technologies®, Electrothermal®, Environmental Express™, Ismatec®, Jenway®, Kinesis®, Masterflex®, PCRmax®, Stuart®, Techne®, TELOS®, Traceable®, Vaplock™ and a wide range of private-label products.
Follow us on Twitter and Facebook
® Cole-Parmer Ltd, 2018.
Editorial enquiries:
Pranika Sivakumar, Notch Communications
+44 161 457 7230
[email protected]
Popular Categories
- Balances and Scales
- Analytical Balances
- Bench and Floor Scale Bases
- Bench and Floor Scales
- Counting Scales
- Crane and Hanging Scales
- Health Scales
- Indicators for Scales
- Mechanical Balances
- Moisture Balances
- Platform and Pallet Scales
- Production Weighing
- Semi and Micro Analytical Balances
- Toploading Balances
- Weighing Accessories
- Weighing Dishes
- Calibration
- Electrical Products
- Batteries and Battery Testers
- Cable Testers
- Cable Ties
- Clamp Meters
- Electrical Accessories
- Electronics Enclosures
- Function Generators
- Gauss Meters
- Insulation Testers
- LCR Meters
- Megohmmeters
- Motor Rotation Testers
- Multimeters
- Oscilloscopes
- Outlet Strips
- Power Meters and Analyzers
- Power Supplies
- Resistance Testers
- RF Signal Generators
- Static Control Products
- Surge Protectors
- Test Leads and Probes
- Voltage Detectors
- Filtration Products
- Fluid Handling
- Furniture
- Heating and Cooling Equipment
- Laboratory Equipment
- Baths and Circulators
- Burettes (Burets)
- Calorimeters
- Centrifuges
- Chromatography Products
- Desiccators
- Digestion Systems
- Dispensers
- Distillation
- Electrophoresis Products
- Electroporation Products
- Environmental Samplers
- Extraction Apparatus
- Fermentation Products
- Flame Photometers
- Fluid Bed Dryers
- Freeze Dryers
- Glassware Washers
- Histology Products
- Homogenizers and Ultrasonic Processors
- Humidifiers and Dehumidifiers
- Lab Shakers
- Laboratory Evaporators
- Laboratory Sieves and Sieve Shakers
- Leachate Equipment
- Lighting and Illumination
- Magnetic Stirrers and Hot Plates
- Microscopes and Magnification
- Mixers and Blenders
- Particle Counters
- Pipettes and Pipettors
- Refractometers
- Thermal Cyclers
- Titrators
- Ultrasonic Cleaners
- Vacuums
- Water Purification Products
- Laboratory Supplies
- Air Sampling
- Animal Research Products
- Bags
- Beakers
- Books and Reference Materials
- Bottles and Jars
- Calculators
- Carboys
- Chemistry Kits
- Clean Room Stationery
- Containers
- Cuvettes
- Dialysis Products
- Dippers
- Distillation Supplies
- Field Sampling
- Flasks
- Graduated Cylinders
- Ice Buckets and Ice Pans
- Lab Cleaning Products
- Lab Organizers
- Labels and Label Printers
- Laboratory Dishes
- Laboratory Timers and Clocks
- Labware
- Microbiology Products
- Microplate Products
- Mortars and Pestles
- Parts and Accessories
- Racks
- Sampling Wipes
- Shippers
- Syringes
- Tubes
- Vials
- Watch Glasses
- Material Handling and Storage
- Material Testing and Physical Testing
- Pressure and Vacuum Instruments
- Process Controllers
- Pumps
- Safety Products
- Air Quality Monitors
- Arc Flash Products
- Combustion Analyzers
- Eyewash
- Eyewear and Safety Glasses
- Floor Mats and Matting
- Gas Detectors
- Gloves
- Hearing Protection
- Lockout and Tagout Product
- Personal Monitor Badges
- Radiation Detectors
- Refrigerant Products
- Respirators and Masks
- Safety Apparel
- Safety Shields
- Safety Signs and Labels
- Solvent Safety
- Sound Meters
- Spill Control
- Temperature Measurement Products
- Test and Measurement Products
- Water Quality Products
- Colorimeters
- Colorimetry Reagents
- Conductivity, Resistivity, and TDS Meters
- Dissolved Oxygen Meters
- Dye Tracers
- Electrodes
- Flocculation Testers
- Multiparameter Meters
- pH Buffers
- pH Meters
- Turbidity Accessories
- Turbidity Meters
- Water Quality Accessories
- Water Quality Standards, and Solutions
- Water Quality Test Strips