Cole-Parmer® TELOS® neo™ MicroPlate™ SPE Populated 96-Well Microplates
50 µL elution volume for minimal sample dilution and reduced evaporation time
  • Versatile 96-well format for small volume biological fluid samples
  • Optimized well geometry for 5 and 10 mg sorbent masses
  • Use with vacuum or positive pressure
The internal diameter, frit design, and sorbent material of the Cole-Parmer® TELOS® neo™ MicroPlate™ 96-well plates allow for maximum recovery with minimal sample dilution and reduced evaporation time. A well packed with 5 mg sorbent allows analytes to be eluted in as little as 50 µL.
Populated plate comes with 96 wells already in place. Modular design lets you simply remove any unwanted wells and add sealing strips. Full or partially populated plates can be processed using either vacuum or positive pressure.
33,800.00 - ₹49,400.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sorbent Applications Bed Mass (mg)
Availability Pricing
COMPARE
PAX Mixed-mode strong anion exchange 5
₹42,600.00INR / Each
COMPARE
WAX Mixed-mode weak anion exchange 10
₹44,300.00INR / Each
COMPARE
WCX Mixed-mode weak cation exchange 5
₹42,600.00INR / Each
COMPARE
WCX Mixed-mode weak cation exchange 10
₹49,400.00INR / Each
COMPARE
PCX Mixed-mode strong cation exchange 10
₹40,700.00INR / Each
COMPARE
PRP Nonpolar 10
₹42,700.00INR / Each
COMPARE
PCR Mixed-mode strong cation exchange 5
₹34,900.00INR / Each
COMPARE
WAX Mixed-mode weak anion exchange 5
₹41,000.00INR / Each
COMPARE
PRP Nonpolar 5
₹33,800.00INR / Each
Showing 1- 9 of 9