Cole-Parmer TELOS® PAX MicroPlate™ 96-well Plate, populated, 5mg / 1mL, 1 / pk
Representative image only.
Cole-Parmer TELOS® PAX MicroPlate™ 96-well Plate, populated, 5mg / 1mL, 1 / pk
Cole-Parmer | Item: 0647402
50 µL elution volume for minimal sample dilution and reduced evaporation time
₹47,800.00INR / Each
₹47,800.00INR / Each
Buy It Together
Current Item
Cole-Parmer TELOS®  PAX MicroPlate™ 96-well Plate, populated, 5mg / 1mL, 1 / pk
Cole-Parmer TELOS® PAX MicroPlate™ 96-well Plate, populated, 5mg / 1mL, 1 / pk
0647402
₹47,800.00INR / Each
+
Cole-Parmer TELOS® MicroPlate Sealing Strips, 24 / pk
0647411
₹5,700.00INR / Pkg of 24
=
Total Price 53500.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Versatile 96-well format for small volume biological fluid samples
  • Optimized well geometry for 5 and 10 mg sorbent masses
  • Use with vacuum or positive pressure
  • Applications: Mixed-mode strong anion exchange sorbent for acidic analyte extraction
More About this Item
The internal diameter, frit design, and sorbent material of the Cole-Parmer® TELOS® neo™ MicroPlate™ 96-well plates allow for maximum recovery with minimal sample dilution and reduced evaporation time. A well packed with 5 mg sorbent allows analytes to be eluted in as little as 50 µL.
Populated plate comes with 96 wells already in place. Modular design lets you simply remove any unwanted wells and add sealing strips. Full or partially populated plates can be processed using either vacuum or positive pressure.
Populated plate comes with 96 wells already in place. Modular design lets you simply remove any unwanted wells and add sealing strips.
Specifications
  • Bed Mass (mg)5
  • Column Particle Size (µm)60
  • Bottom TypePolymer based
  • ApplicationsMixed-mode strong anion exchange
  • SorbentPAX
  • DescriptionPAX MicroPlate SPE Microplate, populated, 5 mg sorbent; 1/ea