Digi-Sense Touch-Screen Recorder
3/6/2017
Digi-Sense Touch-Screen Recorder from Cole-Parmer Provides Virtually Unlimited Datalogging
March 6, 2017, Vernon Hills, Ill — The Digi-Sense Touch Screen Recorder, from Cole-Parmer, is a tablet-based instrument equipped with 8GB of onboard memory for virtually unlimited datalogging capacity. An intuitive touch-screen display makes the recorder easy to use and view. It is ideal for monitoring and logging environmental conditions in food, pharmaceutical, laboratory, greenhouse, museum or other sensitive areas.
The touch-screen recorder offers many user-friendly features. It measures, displays, and logs temperature, relative humidity and dew point conditions. The measured values are simultaneously displayed both graphically and digitally on a color touch-screen display. Operation is simple right out of the box. The resident operating software self-boots by pushing the on/off button. The touch-screen menu makes it easy to set up data logging attributes including sample frequency, sample size, alarm settings, and units of measure. The recorder can be placed in real-time mode or set to start logging at some future time. It also comes with NIST traceable calibration.
To learn more, go to coleparmer.com/c/dataloggers
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Ismatec®, Oakton®, Digi-Sense™, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 1-800-323-4340. International customers, call 1-847-549-7600. Visit us at www.coleparmer.com.
Download jpg image