Traceable® Temperature/Humidity Touchscreen Recorder
8GB on board memory provides virtually unlimited data logging capacity
  • Measures, logs, and displays temperature, humidity, and dew point
  • Removable sensor for local or remote measurements using extension cable included
  • Resident self-booting software, no installation necessary
  • Stored data downloads to computer via USB cable provided in CSV format
  • Wall mount or desk mount operation
  • Includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory
This tablet-based instrument measures, displays, and logs temperature, relative humidity and dew point conditions. Simultaneously view measured values both graphically and digitally on a 6" x 3-1/2", high-resolution color touch-screen display. Operation is very simple out of the box; push the on/off button and the resident operating software self-boots. Set up data logging attributes including sample frequency, sample size, alarm settings, and units of measure through an intuitive touchscreen menu. Set the recorder for real-time mode or to start logging at some future time.
Easily wall mount the instrument using included bracket or place it on a horizontal surface using the built-in stand. Plug the temperature/RH sensor directly onto the back of the tablet or remotely up to 6 ft away using an extension cable (included). Add additional extension cables 20250-47 (sold separately) to extend the sensor up to 18 ft from the instrument. A 110 VAC adapter powers the instrument and a 2500 mAH on-board battery provides backup up to six hours in case of power interruption.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Traceable Temperature/Humidity Touchscreen Recorder 2025040 Mfr # 9012TR
COMPARE
Temperature/Humidity Touchscreen Recorder