Zetex High-Temperature Gloves and Finger Cots
  • Material resists up to 1000°F (538°C)
A safe alternative to asbestos, these gloves and finger cots are made of a durable cotton-backed, 11-ounce wool liner. Liner is sewn at the fingertips and double stitched with aramid thread throughout for strength and durability. One size fits all. Made in the USA. Compliance: European EN407 industrial standard
Note: The material resists up to 1000°F (538°C); operating temperature is 700°F (371°C) for longer exposure. These temperatures are dependent on duration, application, and environment. Actual use temperature may be lower.
19,900.00 - ₹28,700.00INR / Pair of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Newtex Industries ZG1 High-Temp Gloves, 1000 F, 14"L, One-Size; One Pair WW-86364-00 Mfr # 2100006
COMPARE
High-Temp Gloves, 1000 F, 14\"L, One-Size; One Pair
₹19,900.00INR / Pair of 1
Newtex Industries Zetex High-Temp Gloves, Aluminized-Back, 1000F, 14"L, One-Size; One Pair WW-86364-20 Mfr # 2100019
COMPARE
High-Temp Gloves, Aluminized-Back, 1000F, 14\"L, One-Size; One Pair
₹28,700.00INR / Pair of 1