YSI Conductivity Calibration Solutions
Referenced to primary standard solutions according to OIML recommendation 56
  • Values are accurate to ±0.25 to ±0.5%
Each 32-oz bottle comes with NIST-traceable calibration report supplied by manufacturer.
24,000.00 - ₹34,000.00INR / Quart of 1
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Conductivity Value (µS) Description
Availability Pricing
YSI 3161 Conductivity calibration standard, 1000 µmhos 0148254 Mfr # 065270
COMPARE
1000 Conductivity calibration standard, 1000 µmhos
₹24,900.00INR / Quart of 1
YSI 3163 conductivity calibration standard, 10,000 µS 0148256 Mfr # 065272
COMPARE
10000 conductivity calibration standard, 10,000 µS
₹24,000.00INR / Quart of 1
YSI 3165 conductivity calibration standard, 100,000 µS 0148258 Mfr # 065274
COMPARE
100000 conductivity calibration standard, 100,000 µS
₹34,000.00INR / Quart of 1