YSI 910 COD Reactor, Test Kits, and Colorimeter
US EPA-approved COD test method
  • Premeasured COD reagents cover three test ranges
  • 430 nm single-wavelength colorimeter is specific to COD testing
  • Digest up to 24 samples simultaneously with thermal reactor
Testing requires the purchase of all three components: a COD colorimeter, a thermal reactor, and COD test reagents.
479,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
YSI Thermoreactor, 24 Vials, 115 V 9955025 Mfr # 1P22-2Y
COMPARE
Thermoreactor, 24 Vials, 115 V
₹479,000.00INR / Each