WTW Water Analysis Accessories

Optimize water analysis
  • Choose from a variety of accessories
Replacement electrodes and probes, calibration standards, cables, and other parts for use with WTW instruments.
61,300.00 - ₹180,000.00INR / Each

10 variations of this product are available.

See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
WTW 301995 KLE 325 Conductivity Cell ( ProfiLine Meters) 1970781 Mfr # 301995
COMPARE
KLE 325 Conductivity Cell ( ProfiLine Meters)
₹180,000.00INR / Each
WTW 201545 Replacement standard DO probe, 3-meter cable 5310550 Mfr # 201545
COMPARE
Replacement standard DO probe, 3-meter cable
Call For Price
WTW 202706 Heavy-duty DO probe maintenance kit, for CellOx probes 5310553 Mfr # 202706
COMPARE
Heavy-duty DO probe maintenance kit, for CellOx probes
Call For Price
WTW 201425 Replacement auto-stir probe 9 ½" x 9" x 4 ½" 5310650 Mfr # 201425-CR
COMPARE
Replacement auto-stir probe 9 ½" x 9" x 4 ½"
Call For Price
WTW 202710 Replacement membrane kit for auto-stir probe 53106-50 5310651 Mfr # 202710
COMPARE
Replacement membrane kit for auto-stir probe 53106-50
Call For Price
WTW 103-635 Replacement Waterproof Combination pH Electrode 5890055 Mfr # 103635
COMPARE
Replacement Waterproof Combination pH Electrode
₹96,900.00INR / Each
WTW 103631 SenTix 21 pH Electrode, Epoxy Body, Gel Fill; 1 m Cable 5890850 Mfr # 103631
COMPARE
SenTix 21 pH Electrode, Epoxy Body, Gel Fill; 1 m Cable
₹61,300.00INR / Each
WTW 103635 SenTix 41 pH/Temp Electrode, Epoxy Body; 1 m Cable 5890855 Mfr # 103635
COMPARE
SenTix 41 pH/Temp Electrode, Epoxy Body; 1 m Cable
₹96,900.00INR / Each
WTW 103651 pH Electrode Sentix 51 1-m cable DIN connector 5890857 Mfr # 103651
COMPARE
pH Electrode Sentix 51 1-m cable DIN connector
₹124,000.00INR / Each
WTW 103636 Combination pH electrode, gel filled with temperature compensation and 10-ft cable 9957853 Mfr # 103636
COMPARE
Combination pH electrode, gel filled with temperature compensation and 10-ft cable
₹139,000.00INR / Each
Showing 1- 10 of 10

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat