Wilmad-LabGlass Thin Wall Precision NMR Sample Tubes
Maximize SNR with precision NMR tubes that have minimal paramagnetic impurities
  • NMR tubes are constructed of ASTM E438 Type 1 Class B borosilicate glass
  • Precision tubes have 3-fold less paramagnetic contamination comparing to the Economy tube
  • Rated as hydrolytic class 1— inner surface is resistant to strong acid and base at ambient temperature
  • 100% visual and physical inspection ensures quality, including physical dimensions and surface defects
These Wilmad-LabGlass thin wall precision NMR sample tubes are manufactured in a state-of-the-art ISO 9001:2015 USA facility using a unique precision shrinking and grinding process to shape the inner surface with maximized filling factor. To maximize SNR, the precision NMR tubes have minimal paramagnetic impurities that would impact shimming. Tight ID and OD tolerance as small as 0.0065 mm accommodates Wilmad inserts. Tubes are rated as hydrolytic class 1, meaning the inner surface is resistant to strong acid and base at ambient temperature. All tubes are 100% inspected with multiple NIST traceable gauges and optical surface defect checks to ensure accuracy and consistency. Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.
10,100.00 - ₹59,400.00INR / Pkg of 5
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Special Features Tube Length (mm) Description
Availability Pricing
Wilmad-LabGlass Thin Wall Precision NMR Sample Tube, 7" L, 500 MHz; 5/Pk WW-83999-50 Mfr # 528-PP-7-5
COMPARE
500 MHz 178 Thin Wall Precision NMR Sample Tube, 7" L, 500 MHz; 5/Pk
₹26,900.00INR / Pkg of 5
Wilmad-LabGlass Thin Wall Precision NMR Sample Tube, 7" L, 300 MHz; 5/Pk WW-83999-54 Mfr # 507-PP-7-5
COMPARE
300 MHz 178 Thin Wall Precision NMR Sample Tube, 7" L, 300 MHz; 5/Pk
₹18,700.00INR / Pkg of 5
Wilmad-LabGlass Thin Wall Precision NMR Sample Tube, 7" L, 200 MHz; 5/Pk WW-83999-77 Mfr # 506-PP-7-5
COMPARE
200 MHz 178 Thin Wall Precision NMR Sample Tube, 7" L, 200 MHz; 5/Pk
₹10,100.00INR / Pkg of 5
Wilmad-LabGlass Thin Wall Precision NMR Sample Tube, 8" L, 200 MHz; 5/Pk WW-83999-78 Mfr # 506-PP-8-5
COMPARE
200 MHz 203.2 Thin Wall Precision NMR Sample Tube, 8" L, 200 MHz; 5/Pk
₹11,900.00INR / Pkg of 5
Wilmad-LabGlass Thin Wall Precision NMR Sample Tube, 7" L, 350 MHz; 5/Pk WW-83999-79 Mfr # 526-PP-7-5
COMPARE
350 MHz 178 Thin Wall Precision NMR Sample Tube, 7" L, 350 MHz; 5/Pk
₹16,700.00INR / Pkg of 5
Wilmad-LabGlass Thin Wall Precision NMR Sample Tube, 8" L, 350 MHz; 5/Pk WW-83999-80 Mfr # 526-PP-8-5
COMPARE
350 MHz 203.2 Thin Wall Precision NMR Sample Tube, 8" L, 350 MHz; 5/Pk
₹18,500.00INR / Pkg of 5
Wilmad-LabGlass Thin Wall Precision NMR Sample Tube, 7" L, 400 MHz; 5/Pk WW-83999-81 Mfr # 527-PP-7-5
COMPARE
400 MHz 178 Thin Wall Precision NMR Sample Tube, 7" L, 400 MHz; 5/Pk
₹17,600.00INR / Pkg of 5
Wilmad-LabGlass Thin Wall Precision NMR Sample Tube, 8" L, 400 MHz; 5/Pk WW-83999-82 Mfr # 527-PP-8-5
COMPARE
400 MHz 203.2 Thin Wall Precision NMR Sample Tube, 8" L, 400 MHz; 5/Pk
₹19,600.00INR / Pkg of 5
Wilmad-LabGlass Thin Wall Precision NMR Sample Tube, 7" L, 600 MHz; 5/Pk WW-83999-83 Mfr # 535-PP-7-5
COMPARE
600 MHz 178 Thin Wall Precision NMR Sample Tube, 7" L, 600 MHz; 5/Pk
₹29,900.00INR / Pkg of 5
Wilmad-LabGlass Thin Wall Precision NMR Sample Tube, 8" L, 600 MHz; 5/Pk WW-83999-84 Mfr # 535-PP-8-5
COMPARE
600 MHz 203.2 Thin Wall Precision NMR Sample Tube, 8" L, 600 MHz; 5/Pk
₹33,000.00INR / Pkg of 5
Wilmad-LabGlass Thin Wall Precision NMR Sample Tube, 7" L, 800 MHz; 5/Pk WW-83999-85 Mfr # 541-PP-7-5
COMPARE
800 MHz 178 Thin Wall Precision NMR Sample Tube, 7" L, 800 MHz; 5/Pk
₹46,000.00INR / Pkg of 5
Wilmad-LabGlass Thin Wall Precision NMR Sample Tube, 8" L, 800 MHz; 5/Pk WW-83999-86 Mfr # 541-PP-8-5
COMPARE
800 MHz 203.2 Thin Wall Precision NMR Sample Tube, 8" L, 800 MHz; 5/Pk
₹50,200.00INR / Pkg of 5
Wilmad-LabGlass Thin Wall Precision NMR Sample Tube, 7" L, 1000 MHz; 5/Pk WW-83999-87 Mfr # 542-PP-7-5
COMPARE
1000 MHz 178 Thin Wall Precision NMR Sample Tube, 7" L, 1000 MHz; 5/Pk
₹59,400.00INR / Pkg of 5
Wilmad-LabGlass Thin Wall Precision NMR Sample Tube, 8" L, 500 MHz; 5/Pk WW-83999-89 Mfr # 528-PP-8-5
COMPARE
500 MHz 203.2 Thin Wall Precision NMR Sample Tube, 8" L, 500 MHz; 5/Pk
₹24,300.00INR / Pkg of 5
Showing 1- 14 of 14