Wilmad-LabGlass Economy NMR Sample Tubes
Guaranteed to fit tightly in any spinner turbine and have zero NMR background
  • NMR tubes are constructed of ASTM E438 Type 1 Class B borosilicate glass
  • Camber and concentricity values listed are the Total Indicator Reading (TIR)
  • 100% visual and physical inspection ensures quality, including physical dimensions and surface defects
These Wilmad-LabGlass NMR economy sample tubes are designed for use in all NMR spectrometers and are guaranteed to fit tightly in any spinner turbine and have zero NMR background. Each 5mm tube meets precise wall thickness to ensure they fit tightly in spinner turbine, these tubes have the best OD tolerance in the industry and a 30% thicker wall than other products. All tubes are made from borosilicate glass and manufactured in an ISO 9001:2015 USA facility. Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.
5,500.00 - ₹49,700.00INR / Pkg of 5
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Special Features Tube Length (mm) Tube Outside Diameter (OD) (mm)
Availability Pricing
SP Wilmad NMR Economy Sample Tube, Borosilicate Glass, 4.947 mm OD x 7" L, 100 MHz; 5/Pk 8399964 Mfr # WG-1206-7-5
COMPARE
100 MHz 177.8 4.947
₹8,900.00INR / Pkg of 5
SP Wilmad NMR Economy Sample Tube, Borosilicate Glass, 4.947 mm OD x 7" L, 200 MHz; 5/Pk 8399965 Mfr # WG-1208-7-5
COMPARE
200 MHz 177.8 4.947
₹9,900.00INR / Pkg of 5
SP Wilmad NMR Economy Sample Tube, Borosilicate Glass, 4.947 mm OD x 7" L, 300 MHz; 5/Pk 8399966 Mfr # WG-1226-7-5
COMPARE
300 MHz 177.8 4.947
₹12,000.00INR / Pkg of 5
SP Wilmad NMR Economy Sample Tube, Borosilicate Glass, 4.947 mm OD x 8" L, 300 MHz; 5/Pk 8399967 Mfr # WG-1226-8-5
COMPARE
300 MHz 203.2 4.947
₹16,900.00INR / Pkg of 5
SP Wilmad NMR Economy Sample Tube, Borosilicate Glass, 4.947 mm OD x 7" L, 400 MHz; 5/Pk 8399968 Mfr # WG-1228-7-5
COMPARE
400 MHz 177.8 4.947
₹13,300.00INR / Pkg of 5
SP Wilmad NMR Economy Sample Tube, Borosilicate Glass, 4.947 mm OD x 8" L, 400 MHz; 5/Pk 8399969 Mfr # WG-1228-8-5
COMPARE
400 MHz 203.2 4.947
₹17,100.00INR / Pkg of 5
SP Wilmad NMR Economy Sample Tube, Borosilicate Glass, 4.947 mm OD x 7" L, 500 MHz; 5/Pk 8399970 Mfr # WG-1235-7-5
COMPARE
500 MHz 177.8 4.947
₹18,700.00INR / Pkg of 5
SP Wilmad NMR Economy Sample Tube, Borosilicate Glass, 4.947 mm OD x 8" L, 500 MHz; 5/Pk 8399971 Mfr # WG-1235-8-5
COMPARE
500 MHz 203.2 4.947
₹23,600.00INR / Pkg of 5
SP Wilmad NMR Economy Sample Tube, Borosilicate Glass, 4.947 mm OD x 7" L, 600 MHz; 5/Pk 8399972 Mfr # WG-1241-7-5
COMPARE
600 MHz 177.8 4.947
₹21,500.00INR / Pkg of 5
SP Wilmad NMR Economy Sample Tube, Borosilicate Glass, 4.947 mm OD x 8" L, 600 MHz; 5/Pk 8399973 Mfr # WG-1241-8-5
COMPARE
600 MHz 203.2 4.947
₹26,700.00INR / Pkg of 5
SP Wilmad NMR Economy Sample Tube, Borosilicate Glass, 4.947 mm OD x 7" L, 700 MHz; 5/Pk 8399974 Mfr # WG-1242-7-5
COMPARE
700 MHz 177.8 4.947
₹49,700.00INR / Pkg of 5
SP Wilmad NMR Economy Sample Tube, Borosilicate Glass, 4.94665 mm OD x 7" L, 100 MHz; 5/Pk 8399975 Mfr # WG-5MM-ECONOMY-7-5
COMPARE
100 MHz 177.8 4.94665
₹5,500.00INR / Pkg of 5
SP Wilmad NMR Economy Sample Tube, Borosilicate Glass, 4.94665 mm OD x 8" L, 100 MHz; 5/Pk 8399976 Mfr # WG-5MM-ECONOMY-8-5
COMPARE
100 MHz 203.2 4.94665
₹6,900.00INR / Pkg of 5
Showing 1- 13 of 13