Wildco Indestructible Aquatic Tri-Net
Extremely durable, all-purpose net for heavily weeded and overgrown areas
  • Mouth is 12" x 12" x 12"
  • Net is 6" deep
  • Handle is 52" L x 1-1/4" dia
Triangular frame provides extreme rigidity needed for digging. The net has a full muslin shroud to protect the mesh and a Dacron® wear band along the bottom edge. Stainless steel net ring and removable, hardwood handle will provide years of use.
52,600.00 - ₹54,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Material Sample Volume
Availability Pricing
Wildco 425-T12 Aquatic Tri-Net, multifilament material, 800/900 um 0549150 Mfr # 425-T12
COMPARE
Aquatic Tri-Net, multifilament material, 800/900 um Stainless Steel 800/900 um
₹54,100.00INR / Each
Wildco 425-T52 Aquatic Tri-Net, nylon material, 500 um 0549152 Mfr # 425-T52
COMPARE
Aquatic Tri-Net, nylon material, 500 um Stainless Steel 500 um
₹52,600.00INR / Each