Wildco D-Frame Aquatic Dip Nets
Heavy-duty, canvas bottom bag for open and lightly weeded areas
  • Mouth is 12" W x 10" tall
  • Net is 22" Deep
  • Handle is 52" L x 1-1/4" dia
Stainless steel net ring and removable hardwood handle will provide years of use. Canvas shroud on bottom of net protects it from snags while still allowing clear view of contents.
52,400.00 - ₹65,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Material
Availability Pricing
Wildco 425-D12 D-frame Multifilment 800/900 um Net 0549130 Mfr # 425-D12
COMPARE
D-frame Multifilment 800/900 um Net Stainless Steel
₹52,400.00INR / Each
Wildco 425-D52 D-frame Multifilment 500 um (EPA) Net 0549132 Mfr # 425-D52
COMPARE
D-frame Multifilment 500 um (EPA) Net Stainless Steel
₹65,200.00INR / Each