Wildco Aquatic Kick Nets
Muslin shroud on the bottom provides protection and viewing ability from above
  • Mouth is 18"W x 9" tall
  • Net is 10" deep
  • Two-piece handle is 60" L x 1-1/4" dia
Use these kick nets in stream beds. As one person holds the net, another kicks the stream bed or runs their fingers through the substrate to drive invertebrates into the net. Stainless steel net ring and removable, two-piece hardwood handle will provide years of use.
68,000.00 - ₹79,300.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Material
Availability Pricing
Wildco 425-K13 Aquatic Kick Net, multifilament material, 800-900 um 0549140 Mfr # 425-K13
COMPARE
Aquatic Kick Net, multifilament material, 800-900 um Stainless Steel
₹68,000.00INR / Each
Wildco 425-K53 Aquatic Kick Net, nylon material, 500 um 0549142 Mfr # 425-K53
COMPARE
Aquatic Kick Net, nylon material, 500 um Stainless Steel
₹79,300.00INR / Each