WHIRL-PAK® Sterile Sample Bags with Hydrated Sampling Sponge
Make environmental surface sampling faster and easier
  • Write-on label makes identifying samples easy and convenient
  • Each bag contains a polyurethane sponge, premoistened with HiCap™ neutralizing broth
  • Terminally gamma processed to guarantee sterility
Collection broths play a critical role in the success of an environmental monitoring program. HiCap™ neutralizing broth is the first collection broth developed specifically for use in the food and beverage industry as an alternative to traditional collection broths. The polyurethane sponges included with each bag are nontoxic and do not contain biocides. Unlike the “wet” cellulose manufacturing process, there is no need to include biocides to control microbial growth during the polymerization process.
97,000.00 - ₹131,000.00INR / Box of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Whirl-Pak B01590WA Hydrated PolySponge™ Sampling Bag with Sampling Sponge, 18 oz, Sterile; 100/Bx 0064991 Mfr # B01590
COMPARE
Sampling Bag with Sampling Sponge, 18 oz, Sterile; 100/Bx
₹97,000.00INR / Box of 100
Whirl-Pak B01592WA Hydrated PolyProbe™ Sampling Bag with Sampling Sponge and 8"L Probe, 24 oz, Sterile; 100/Bx 0064993 Mfr # B01592
COMPARE
Sampling Bag with Sampling Sponge and 8"L Probe, 24 oz, Sterile; 100/Bx
₹131,000.00INR / Box of 100