WHEATON® Tubular Serum Vials, European Blowback, ISO 8362-1
Shrink-wrapped modules reduce particulate contamination
  • Made from Type I borosilicate glass tubing
  • Vials conform to requirements outlined in ISO 8362 EP
These WHEATON® tubular serum vials with European blowback, conform to ISO 8362 EP standards, and are constructed of Type I borosilicate glass. The blowback feature in the neck and specially designed bottom radius for added strength are important for lyophilization applications. Vials are packaged in shrink wrapped trays to greatly reduce particulate contamination.
26,700.00 - ₹47,700.00INR / Case of 196
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Outside Diameter (mm) Special Features Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) Tubular Serum Vials, European Blowback, ISO 8362-1, Clear Glass, 2R, 4 mL; 196/CS 0891628 Mfr # WEBB02R-T-EP
COMPARE
16 Size 2R 4
₹46,300.00INR / Case of 196
DWK Life Sciences (Wheaton) Tubular Serum Vials, European Blowback, ISO 8362-1, Clear Glass, 6R, 10 mL; 100/CS 0891629 Mfr # WEBB06R-T-EP
COMPARE
22 Size 6R 10
₹26,700.00INR / Case of 100
DWK Life Sciences (Wheaton) Tubular Serum Vials, European Blowback, ISO 8362-1, Clear Glass, 10R, 13.5 mL; 150/CS 0891630 Mfr # WEBB10R-T-EP
COMPARE
24 Size 10R 13.5
₹47,700.00INR / Case of 150