Wheaton Socorex Calibrex solutae 530 Bottletop Dispensers
Available in three volume ranges to cover your dispensing needs
  • Built to be autoclaved fully assembled at 250°F (121°C) for 20 minutes
  • Integrated calibration system for easy in-house adjustments
  • Chemically inert and transparent fluid path
The solutae 530 model has a PFA-coated glass plunger for trouble-free dispensing of salt solutions, and both weak and strong acids or bases. A built-in calibration system, hidden under the cap, allows for simple adjustment to accurately accommodate solutions due to varying viscosity or temperature. Two volume adjustment knobs are included—use the premounted slider when changing dispensing volumes frequently or the screw-type version for a more permanent setting without the risk of accidental changes.
174,000.00 - ₹277,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Division (mL) Max Volume (mL) Min Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) Calibrex 530 Socorex solutae Bottletop Dispenser; 2.5 to 25 mL WW-07914-56 Mfr # W844102
COMPARE
0.5 25 2.5
₹174,000.00INR / Each
DWK Life Sciences (Wheaton) Calibrex 530 Socorex solutae Bottletop Dispenser; 5 to 50 mL WW-07914-57 Mfr # W844104
COMPARE
1 50 5
₹185,000.00INR / Each
DWK Life Sciences (Wheaton) Calibrex 530 solutae Bottletop Dispenser; 10 to 100 mL WW-07914-58 Mfr # W844106
COMPARE
1 100 10
₹277,000.00INR / Each