WHEATON® Serum Vials, US Blowback, EP 3.2.1
Special design adds strength for freeze-drying applications
  • Made from Type I borosilicate glass tubing
  • Vials conform to the hydrolytic resistance requirements outlined in USP <660> and EP 3.2.1
  • Shrink-wrapped modules reduce particulate contamination
These WHEATON® serum vials with US blowback are constructed of Type I borosilicate glass, and available in clear or amber colors. The blowback feature in the neck and specially designed bottom radius for added strength are important for lyophilization applications. Vials are packaged in shrink wrapped trays to greatly reduce particulate contamination.
26,700.00 - ₹60,300.00INR / Case of 225
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Outside Diameter (mm) Tube Color Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) Serum Vials, US Blowback, EP 3.2.1, Clear Glass, 2 mL; 225/CS 0891620 Mfr # WUBB2MLC-T-EP
COMPARE
14.75 Clear 2
₹46,300.00INR / Case of 225
DWK Life Sciences (Wheaton) Serum Vials, US Blowback, EP 3.2.1, Amber Glass, 2 mL; 225/CS 0891621 Mfr # WUBB2MLA-T-EP
COMPARE
14.75 Amber 2
₹60,300.00INR / Case of 225
DWK Life Sciences (Wheaton) Serum Vials, US Blowback, EP 3.2.1, Clear Glass, 3 mL; 169/CS 0891622 Mfr # WUBB3MLC-T-EP
COMPARE
16.75 Clear 3
₹35,700.00INR / Case of 169
DWK Life Sciences (Wheaton) Serum Vials, US Blowback, EP 3.2.1, Amber Glass, 3 mL; 169/CS 0891623 Mfr # WUBB3MLA-T-EP
COMPARE
16.75 Amber 3
₹43,700.00INR / Case of 169
DWK Life Sciences (Wheaton) Serum Vials, US Blowback, EP 3.2.1, Clear Glass, 5 mL; 100/CS 0891624 Mfr # WUBB5MLC-T-EP
COMPARE
21.75 Clear 5
₹26,700.00INR / Case of 100
DWK Life Sciences (Wheaton) Serum Vials, US Blowback, EP 3.2.1, Amber Glass, 5 mL; 100/CS 0891625 Mfr # WUBB5MLA-T-EP
COMPARE
21.75 Amber 5
₹31,000.00INR / Case of 100
DWK Life Sciences (Wheaton) Serum Vials, US Blowback, EP 3.2.1, Clear Glass, 10 mL; 150/CS 0891626 Mfr # WUBB10MLC-T-EP
COMPARE
23.75 Clear 10
₹47,700.00INR / Case of 150
DWK Life Sciences (Wheaton) Serum Vials, US Blowback, EP 3.2.1, Amber Glass, 10 mL; 150/CS 0891627 Mfr # WUBB10MLA-T-EP
COMPARE
23.75 Amber 10
₹57,800.00INR / Case of 150
Showing 1- 8 of 8