WHEATON Serum Tubing Vials
Special design adds strength for freeze drying applications
  • Made from low extractable Type 1 borosilicate glass
The blowback feature in the neck and specially designed bottom radius for added strength
are important for lyophilization applications. Vials are packaged in shrink wrapped modules
to greatly reduce particulate contamination. Available in clear or amber glass. Clear vials
also conform to ASTM Type I and Class A requirements.
27,100.00 - ₹38,000.00INR / Case of 144
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Tube Color Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) 223685 Serum Vials, Clear Type I Glass, 5 mL; 144/Cs 0891602 Mfr # 223685
COMPARE
1 37/64 Clear 5
₹27,100.00INR / Case of 144
DWK Life Sciences (Wheaton) Serum Vials, Clear Type I Glass, 20 mL; 120/Cs 0891604 Mfr # 223687
COMPARE
2 9/32 Clear 20
₹38,000.00INR / Case of 120