WHEATON® LAB FILE® Sample Vials
Protects light sensitive products
  • PTFE/14B rubber-lined black phenolic screw caps ideal for organic samples
  • Improved chemical compatibility
Lab Files® vials are shrink wrapped in partioned trays to protect vials and their contents. Clear vials are converted from Type I borosilicate glass tubing. Caps are rubber lined and are attached to vials to help maintain cleanliness.
24,500.00 - ₹33,300.00INR / Case of 144
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Diameter (mm) Tube Color Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) 224882 LAB FILE® Sample Vials, Clear Glass with Rubber-Lined Cap, 4 mL; 144/Cs 0891984 Mfr # 224882
COMPARE
13 Clear 4
₹24,500.00INR / Case of 144
DWK Life Sciences (Wheaton) 224884 LAB FILE® Sample Vials, Clear Glass with Rubber-Lined Cap, 8 mL; 144/Cs 0891986 Mfr # 224884
COMPARE
15 Clear 8
₹28,500.00INR / Case of 144
DWK Life Sciences (Wheaton) 224885 LAB FILE® Sample Vials, Clear Glass with Rubber-Lined Cap, 12 mL; 144/Cs 0891988 Mfr # 224885
COMPARE
15 Clear 12
₹33,300.00INR / Case of 144