WHEATON® Jaw Sets for High-Performance Crimping Tools
Interchangeable jaw sets save you money
  • Compatible with Wheaton High Performance Crimping Tool (sold separately)
Wheaton High Performance Crimping Tool (sold separately) is designed with inter-changeable jaws, so that you only need one tool and purchase the size crimper and decappers needed for your laboratory. These jaw sets are available as crimper and decappers which fit standard lab vials seals with aluminum or two-part aluminum/steel caps.
147,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Cap Diameter (mm)
Availability Pricing
DWK Life Sciences (Wheaton) W225751 Crimping Jaw Set, 11 mm, for High Performance Crimper 9880015 Mfr # W225751
COMPARE
Crimping jaw set 11
₹147,000.00INR / Each
DWK Life Sciences (Wheaton) W225761 Crimping Jaw Set, 13 mm,  for High Performance Crimper 9880016 Mfr # W225761
COMPARE
Crimping jaw set 13
₹147,000.00INR / Each
DWK Life Sciences (Wheaton) W225771 Crimping Jaw Set, 20 mm, for High Performance Crimper 9880017 Mfr # W225771
COMPARE
Crimping jaw set 20
₹147,000.00INR / Each
DWK Life Sciences (Wheaton) W225752 Decapping Jaw Set, 11 mm,  for High Performance Crimper 9880018 Mfr # W225752
COMPARE
Decapping jaw set 11
₹147,000.00INR / Each
DWK Life Sciences (Wheaton) W225762 Decapping Jaw Set, 13 mm, for High Performance Crimper 9880019 Mfr # W225762
COMPARE
Decapping jaw set 13
₹147,000.00INR / Each
DWK Life Sciences (Wheaton) W225772 Decapping Jaw Set, 20 mm, for High Performance Crimper 9880020 Mfr # W225772
COMPARE
Decapping jaw set 20
₹147,000.00INR / Each
DWK Life Sciences (Wheaton) W225763 Flip-Off Crimping Jaw Set, 13 mm,  for High Performance Crimper 9880021 Mfr # W225763
COMPARE
Flip-off crimping jaw set 13
₹147,000.00INR / Each
DWK Life Sciences (Wheaton) W225773 Flip-Off Crimping Jaw Set, 20 mm, for High Performance Crimper 9880022 Mfr # W225773
COMPARE
Flip-off crimping jaw set 20
₹147,000.00INR / Each
Showing 1- 8 of 8