Wheaton® Headspace Vials
Manufactured from clear Type I borosilicate glass
  • Round shoulder and bottom allows for even heating
  • Accepts 20 mm aluminum seals
Wheaton Headspace Vials feature rounded shoulders and bottoms, allowing for even heating and safer operation at higher pressures. The newly designed top provides a tighter seal with your septa. Vials are manufactured from Type I borosilicate glass and accept 20 mm aluminum seals.
Aluminum seals, stopper and septa are sold separately.
11,100.00 - ₹11,400.00INR / Case of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (mm) Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) Headspace Vials, Clear Type I Glass w/o Seals, 6 mL; 100/Cs 0891701 Mfr # 225277
COMPARE
38 6
₹11,100.00INR / Case of 100
DWK Life Sciences (Wheaton) Headspace Vials, Clear Type I Glass w/o Seals, 10 mL; 100/Cs 0891702 Mfr # 225278
COMPARE
46 10
₹11,400.00INR / Case of 100