Wheaton® CompletePAK® Sterile Vial Kits
Reduce scrap costs—equal number of vials, stoppers, and seals
  • Vial packs have sustained particulate cleaning, depryogenation, and sterilization
  • Ready to use—right out of the box
  • 18-month shelf life from date of processing
CompletePAK® vial kits include everything you need, without scrap costs, and no difference in component lead times. These clear-glass vials come with stoppers and seals—choose the volume, cap size and type, and seal color to suit your needs.
These presterilized vial kits are ready to use right off the shelf. Processing includes particulate cleaning, depryogenation, and sterilization—ideal for drug research, freeze drying/lyophilization, aseptic filling, and large/small molecule packaging.
513,000.00 - ₹519,000.00INR / Case of 210
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap color Cap Type Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) CompletePAK® Sterile Vial Kit, 2 mL, Ultra Pure Stopper, Blue; 210/Cs 0891819 Mfr # W991000-CP
COMPARE
Blue Ultra pure, standard 2
₹513,000.00INR / Case of 210
DWK Life Sciences (Wheaton) CompletePAK® Sterile Vial Kit, 2 mL, Ultra Pure Stopper, Red; 210/Cs 0891845 Mfr # W991003-CP
COMPARE
Red Ultra pure, standard 2
₹519,000.00INR / Case of 210