Wheaton® Autosampler ABC VIALS™ Convenience Kits
Save time during sample preparation and reduce the risk of contamination
  • Vial kits consist of vials with thread cap and septa packaged in a PETE clamshell case
Borosilicate glass vials feature 40% larger openings than standard vials—providing improved sample access and minimizing needle damage. Supplied in easy-order convenience kits—including 100 vials and 100 pre-assembled seals with septa. For microvolume sampling, limited-volume inserts are available separately. Inserts feature bottom springs to cushion needle impact.
15,800.00 - ₹18,000.00INR / Pkg of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Septum Type Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) ABC Vial convenience kits with clear vial and PTFE/silicone septa 9875002 Mfr # W225150-04
COMPARE
ABC Vial convenience kits with clear vial and PTFE/silicone septa PTFE/silicone 1.8
₹18,000.00INR / Pkg of 100
DWK Life Sciences (Wheaton) ABC Vial convenience kits with clear vial and PTFE/red silicone septa 9875091 Mfr # W225154-01
COMPARE
ABC Vial convenience kits with clear vial and PTFE/red silicone septa PTFE/red silicone 1.8
₹15,800.00INR / Pkg of 100
DWK Life Sciences (Wheaton) Autosampler Vial Kit, 1.8 mL, Clear, Write-on, PTFE/Silicone; 100/Cs 9875191 Mfr # W225155-01
COMPARE
Autosampler Vial Kit, 1.8 mL, Clear, Write-on, PTFE/Silicone; 100/Cs PTFE/red silicone 1.8
₹17,000.00INR / Pkg of 100