Watson-Marlow PVC 3-Stop Pump Tubing
Use with any Watson-Marlow multichannel cartridges
  • Stops ensure a perfect fit every time
  • Color-coded stops for easy identification
  • Exhibits high flexibility, durability, and strength
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
PVC tubing has hardness shore of 50A and elongation at break of 409% to offer excellent flexibility and durability for use in peristatic pumps. Stops on tubing are color-coded according to tubing ID and ensure a perfect fit on your pump head. Three stops on the tubing offer two pump segments, doubling the life of the tubing. PVC tubing is available with tubing ID from 0.13 to 2.79 mm.
25,900.00 - ₹48,300.00INR / Pkg of 12
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Coded Stops Tubing ID (mm)
Availability Pricing
Watson-Marlow PVC 3-Stop Pump Tubing, 0.13 mm, Orange/Black; 12/Pk 5013052 Mfr # 981.0013.000
COMPARE
Orange/Black 0.13
₹38,700.00INR / Pkg of 12
Watson-Marlow PVC 3-Stop Pump Tubing, 0.19 mm, Orange/Red; 12/Pk 5013053 Mfr # 981.0019.000
COMPARE
Orange/Red 0.19
₹25,900.00INR / Pkg of 12
Watson-Marlow PVC 3-Stop Pump Tubing, 0.25 mm, Orange/Blue; 12/Pk 5013054 Mfr # 981.0025.000
COMPARE
Orange/Blue 0.25
₹25,900.00INR / Pkg of 12
Watson-Marlow PVC 3-Stop Pump Tubing, 0.38 mm, Orange/Green; 12/Pk 5013055 Mfr # 981.0038.000
COMPARE
Orange/Green 0.38
₹25,900.00INR / Pkg of 12
Watson-Marlow PVC 3-Stop Pump Tubing, 0.50 mm, Orange/Yellow; 12/Pk 5013056 Mfr # 981.0050.000
COMPARE
Orange/Yellow 0.5
₹25,900.00INR / Pkg of 12
Watson-Marlow PVC 3-Stop Pump Tubing, 0.63 mm, Orange/White; 12/Pk 5013057 Mfr # 981.0063.000
COMPARE
Orange/White 0.63
₹25,900.00INR / Pkg of 12
Watson-Marlow PVC 3-Stop Pump Tubing, 0.76 mm, Black/Black; 12/Pk 5013058 Mfr # 981.0076.000
COMPARE
Black/Black 0.76
₹25,900.00INR / Pkg of 12
Watson-Marlow PVC 3-Stop Pump Tubing, 0.88 mm, Orange/Orange; 12/Pk 5013059 Mfr # 981.0088.000
COMPARE
Orange/Orange 0.88
₹25,900.00INR / Pkg of 12
Watson-Marlow PVC 3-Stop Pump Tubing, 1.02 mm, White/White; 12/Pk 5013060 Mfr # 981.0102.000
COMPARE
White/White 1.02
₹48,300.00INR / Pkg of 12
Watson-Marlow PVC 3-Stop Pump Tubing, 1.14 mm, Red/Red; 12/Pk 5013061 Mfr # 981.0114.000
COMPARE
Red/Red 1.14
₹25,900.00INR / Pkg of 12
Watson-Marlow PVC 3-Stop Pump Tubing, 1.29 mm, Grey/Grey; 12/Pk 5013062 Mfr # 981.0129.000
COMPARE
Grey/Grey 1.29
₹25,900.00INR / Pkg of 12
Watson-Marlow PVC 3-Stop Pump Tubing, 1.42 mm, Yellow/Yellow; 12/Pk 5013063 Mfr # 981.0142.000
COMPARE
Yellow/Yellow 1.42
₹25,900.00INR / Pkg of 12
Watson-Marlow PVC 3-Stop Pump Tubing, 1.52 mm, Yellow/Blue; 12/Pk 5013064 Mfr # 981.0152.000
COMPARE
Yellow/Blue 1.52
₹25,900.00INR / Pkg of 12
Watson-Marlow PVC 3-Stop Pump Tubing, 1.65 mm, Blue/Blue; 12/Pk 5013065 Mfr # 981.0165.000
COMPARE
Blue/Blue 1.65
₹25,900.00INR / Pkg of 12
Watson-Marlow PVC 3-Stop Pump Tubing, 1.85 mm, Green/Green; 12/Pk 5013066 Mfr # 981.0185.000
COMPARE
Green/Green 1.85
₹25,900.00INR / Pkg of 12
Watson-Marlow PVC 3-Stop Pump Tubing, 2.05 mm, Purple/Purple; 12/Pk 5013067 Mfr # 981.0205.000
COMPARE
Purple/Purple 2.05
₹25,900.00INR / Pkg of 12
Watson-Marlow PVC 3-Stop Pump Tubing, 2.29 mm, Purple/Black; 12/Pk 5013068 Mfr # 981.0229.000
COMPARE
Purple/Black 2.29
₹25,900.00INR / Pkg of 12
Watson-Marlow PVC 3-Stop Pump Tubing, 2.54 mm, Purple/Orange; 12/Pk 5013069 Mfr # 981.0254.000
COMPARE
Purple/Orange 2.54
₹39,600.00INR / Pkg of 12
Watson-Marlow PVC 3-Stop Pump Tubing, 2.79 mm, Purple/White; 12/Pk 5013070 Mfr # 981.0279.000
COMPARE
Purple/White 2.79
₹25,900.00INR / Pkg of 12
Showing 1- 19 of 19