Watson-Marlow PureWeld® XL High-Performance TPE Tubing
Exceptional durability and accurate flow rates—perfect for the most demanding bioprocessing workflows
  • High-performance TPE Tubing offers excellent chemical resistance
  • Ultra-smooth bore minimizes particle entrapment
  • Fully weldable—complete fluid paths in minutes
  • Meets FDA 21CFR117.1810, USP <661>, USP <788>, USP <85>, and USP Class VI
  • Double-packed in heat-sealed bags in Class 7 cleanroom
  • Each bag contains a label with part number, lot number, use-by date, and tube description
  • Sterilize by gamma irradiation
  • Animal-derived content free (ADCF)
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
  • High-performance TPE Tubing offers excellent chemical resistance
Made from high-purity thermoplastic elastomer (TPE), PureWeld® XL tubing offers long service life and consistent flow accuracy. This durable tubing offers excellent chemical resistance, making it suitable for various bioprocessing fluids. The ultra smooth bore supports precise ultrasonic flow measurement and is designed to minimize spallation for cleaner operation.
47,600.00 - ₹117,000.00INR / Pkg
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (mm) Tubing OD (mm) Wall Thickness (mm)
Availability Pricing
Watson-Marlow PureWeld ® XL TPE Tubing, 1.6mm Bore x 2.4mm Wall, 15 m 0075602 Mfr # 942.0016.024
COMPARE
1.6 6.4 2.4
₹70,200.00INR / Pkg
Watson-Marlow Pureweld ® XL TPE Tubing, 1.6mm Bore X 1.6mm Wall, 15 m 0075603 Mfr # 942.0016.016
COMPARE
1.6 4.8 1.6
₹47,600.00INR / Pkg
Watson-Marlow Pureweld ® XL TPE Tubing, 4.8mm Bore X 1.6mm Wall, 15 m 0075604 Mfr # 942.0048.016
COMPARE
4.8 8 1.6
₹51,400.00INR / Pkg
Watson-Marlow Pureweld ® XL TPE Tubing, 4.8mm bore x 2.4mm Wall, 15 m 0075605 Mfr # 942.0048.024
COMPARE
4.8 9.6 2.4
₹84,000.00INR / Pkg
Watson-Marlow Pureweld ® XL TPE Tubing, 6.4mm Bore X 1.6mm Wall, 15 m 0075606 Mfr # 942.0064.016
COMPARE
6.4 9.6 1.6
₹62,700.00INR / Pkg
Watson-Marlow Pureweld ® XL TPE Tubing, 6.4mm bore x 2.4mm Wall, 15 m 0075607 Mfr # 942.0064.024
COMPARE
6.4 11.2 2.4
₹90,200.00INR / Pkg
Watson-Marlow Pureweld ® XL TPE Tubing, 8.0mm Bore X 1.6mm Wall, 15 m 0075608 Mfr # 942.0080.016
COMPARE
8 11.2 1.6
₹68,900.00INR / Pkg
Watson-Marlow Pureweld ® XL TPE Tubing, 8.0mm bore x 2.4mm Wall, 15 m 0075609 Mfr # 942.0080.024
COMPARE
8 12.8 2.4
₹115,000.00INR / Pkg
Watson-Marlow Pureweld ® XL TPE Tubing, 9.6mm bore x 2.4mm Wall, 15 m 0075610 Mfr # 942.0096.024
COMPARE
9.6 14.4 2.4
₹117,000.00INR / Pkg
Showing 1- 9 of 9