Watson-Marlow Marprene® TPE 3-Stop Pump Tubing
Specially formulated thermoelastic elastomer offers extended service life
  • Stops ensure a perfect fit every time
  • Color-coded stops for easy identification
  • Resistant to a wide range of chemicals
  • Highly resistant to oxidizing agents
  • Meets USDA and FDA 21 CFR 177.2600 standards
  • NSF 51 and NSF 61 listed
  • Opaque tubing offers protection from visible and UV light
  • Low gas permeability
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
The durable Marprene TPE tubing lasts at least 10 times as long as other tubing materials, making it perfect for use in peristaltic pumps. Stops on tubing are color-coded according to tubing ID and ensure a perfect fit on your pump head. Three stops on the tubing offer two pump segments, doubling the life of the tubing. This versatile tubing provides wide chemical compatibility and offers high resistance to oxidizing agents such as ozone, peroxides, and sodium hypochlorite.
55,700.00INR / Pkg of 6
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Coded Stops Tubing ID (mm)
Availability Pricing
Watson-Marlow Marprene® TPE 3-Stop Pump Tubing, 0.25 mm, Orange/Blue; 6/Pk 5013035 Mfr # 979.0025.000
COMPARE
Orange/Blue 0.25
₹55,700.00INR / Pkg of 6
Watson-Marlow Marprene® TPE 3-Stop Pump Tubing, 0.38 mm, Orange/Green; 6/Pk 5013036 Mfr # 979.0038.000
COMPARE
Orange/Green 0.38
₹55,700.00INR / Pkg of 6
Watson-Marlow Marprene® TPE 3-Stop Pump Tubing, 0.50 mm, Orange/Yellow; 6/Pk 5013037 Mfr # 979.0050.000
COMPARE
Orange/Yellow 0.5
₹55,700.00INR / Pkg of 6
Watson-Marlow Marprene® TPE 3-Stop Pump Tubing, 0.63 mm, Orange/White; 6/Pk 5013038 Mfr # 979.0063.000
COMPARE
Orange/White 0.63
₹55,700.00INR / Pkg of 6
Watson-Marlow Marprene® TPE 3-Stop Pump Tubing, 0.76 mm, Black/Black; 6/Pk 5013039 Mfr # 979.0076.000
COMPARE
Black/Black 0.76
₹55,700.00INR / Pkg of 6
Watson-Marlow Marprene® TPE 3-Stop Pump Tubing, 0.88 mm, Orange/Orange; 6/Pk 5013040 Mfr # 979.0088.000
COMPARE
Orange/Orange 0.88
₹55,700.00INR / Pkg of 6
Watson-Marlow Marprene® TPE 3-Stop Pump Tubing, 1.02 mm, White/White; 6/Pk 5013041 Mfr # 979.0102.000
COMPARE
White/White 1.02
₹55,700.00INR / Pkg of 6
Watson-Marlow Marprene® TPE 3-Stop Pump Tubing, 1.14 mm, Red/Red; 6/Pk 5013042 Mfr # 979.0114.000
COMPARE
Red/Red 1.14
₹55,700.00INR / Pkg of 6
Watson-Marlow Marprene® TPE 3-Stop Pump Tubing, 1.29 mm, Grey/Grey; 6/Pk 5013043 Mfr # 979.0129.000
COMPARE
Grey/Grey 1.29
₹55,700.00INR / Pkg of 6
Watson-Marlow Marprene® TPE 3-Stop Pump Tubing, 1.42 mm, Yellow/Yellow; 6/Pk 5013044 Mfr # 979.0142.000
COMPARE
Yellow/Yellow 1.42
₹55,700.00INR / Pkg of 6
Watson-Marlow Marprene® TPE 3-Stop Pump Tubing, 1.52 mm, Yellow/Blue; 6/Pk 5013045 Mfr # 979.0152.000
COMPARE
Yellow/Blue 1.52
₹55,700.00INR / Pkg of 6
Watson-Marlow Marprene® TPE 3-Stop Pump Tubing, 1.65 mm, Blue/Blue; 6/Pk 5013046 Mfr # 979.0165.000
COMPARE
Blue/Blue 1.65
₹55,700.00INR / Pkg of 6
Watson-Marlow Marprene® TPE 3-Stop Pump Tubing, 1.85 mm, Green/Green; 6/Pk 5013047 Mfr # 979.0185.000
COMPARE
Green/Green 1.85
₹55,700.00INR / Pkg of 6
Watson-Marlow Marprene® TPE 3-Stop Pump Tubing, 2.05 mm, Purple/Purple; 6/Pk 5013048 Mfr # 979.0205.000
COMPARE
Purple/Purple 2.05
₹55,700.00INR / Pkg of 6
Watson-Marlow Marprene® TPE 3-Stop Pump Tubing, 2.29 mm, Purple/Black; 6/Pk 5013049 Mfr # 979.0229.000
COMPARE
Purple/Black 2.29
₹55,700.00INR / Pkg of 6
Watson-Marlow Marprene® TPE 3-Stop Pump Tubing, 2.54 mm, Purple/Orange; 6/Pk 5013050 Mfr # 979.0254.000
COMPARE
Purple/Orange 2.54
₹55,700.00INR / Pkg of 6
Watson-Marlow Marprene® TPE 3-Stop Pump Tubing, 2.79 mm, Purple/White; 6/Pk 5013051 Mfr # 979.0279.000
COMPARE
Purple/White 2.79
₹55,700.00INR / Pkg of 6
Showing 1- 17 of 17