Water-Level Indicators
Contact of the electrode assembly with the water surface is indicated by an LED and the sound of the audio alert beeper
  • Depth indication is provided by numbered metal tags securely crimped to the cable at intervals of five feet
The brass electrode assembly is 3/8” in diameter and is easily replaceable in the field. The cable is two-conductor, steel and copper plated, with a tensile strength of approximately 300 lbs and a tough plastic type insulating jacket. It is mounted on a small hand-held spool and stand which can be used conveniently to let out or withdraw the cable and electrode assembly.
132,000.00 - ₹294,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Fisher Research Labs WLT100 Water Level Indicator 100FT. 9905010 Mfr # WLT100
COMPARE
Water Level Indicator 100FT.
₹132,000.00INR / Each
Fisher Research Labs WLT200 Water Level Indicator 200FT 9905012 Mfr # WLT200
COMPARE
Water Level Indicator 200FT
₹152,000.00INR / Each
Fisher Research Labs WLT300 Water Level Indicator 300FT 9905014 Mfr # WLT300
COMPARE
Water Level Indicator 300FT
₹212,000.00INR / Each
Fisher Research Labs WLT500 Water Level Indicator 500FT 9905016 Mfr # WLT500
COMPARE
Water Level Indicator 500FT
₹294,000.00INR / Each