Wagner Force Dials
  • Accuracy ±2 graduations <2500 g; ±1 graduation >2500 g
  • Handheld or test stand, mounting holes provided
  • Dual graduations for decimal pounds/grams and ounce/grams.
  • Tension/compression, push and pull from opposite ends
  • Maximum reading memory, maximum reading held automatically; push-button returns pointer to zero
  • Compact size and weight 2.25 in. dial diameter; 10 oz weight
  • Plunger movement, approximately 0.4" (10 mm) full scale deflection
  • Construction, high-impact plastic case, aluminum dial, galvanized steel spring, stainless steel plungers and accessories, brass mechanism chassis and plexiglass dial crystal
The FDK Series force dials feature both decimal lb/gram models as well as equivalent oz/gram models. Now with nine models, they are even more useful in a broad range of push-pull applications up to 60 lb. These units feature improved readability, a variety of accessories and carrying case.
122,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Grads. Description
Availability Pricing
Wagner Instruments FDK-80 Analog Dial Force Gauge; 5 lb (80 oz) 9394949 Mfr # FDK-80
COMPARE
0.25 oz / 25 g Analog Dial Force Gauge; 5 lb (80 oz)
₹122,000.00INR / Each
Wagner Instruments FDK-2 Push/Pull Force Gauge 2 X .020 Lbs Capacity 9394954 Mfr # FDK-2
COMPARE
0.020 lb / 10 g Push/Pull Force Gauge 2 X .020 Lbs Capacity
₹122,000.00INR / Each
Wagner Instruments FDK-5 Push/Pull Force Gauge 5 X .050 Lbs.Capacity 9394956 Mfr # FDK-5
COMPARE
0.050 lb / 25 g Push/Pull Force Gauge 5 X .050 Lbs.Capacity
₹122,000.00INR / Each
Wagner Instruments FDK-10 Push/Pull Force Gauge 10 X .100 Lbs. Capacity 9394957 Mfr # FDK-10
COMPARE
0.100 lb / 50 g Push/Pull Force Gauge 10 X .100 Lbs. Capacity
₹122,000.00INR / Each
Wagner Instruments FDK-20 Push-Pull Force Gauge 20 X .250 Lbs.Capacity 9394958 Mfr # FDK-20
COMPARE
0.250 lb / 100 g Push-Pull Force Gauge 20 X .250 Lbs.Capacity
₹122,000.00INR / Each
Wagner Instruments FDK-40 Push/Pull Force Gauge 40 X .500LBS.Capacity 9394959 Mfr # FDK-40
COMPARE
0.500 lb / 200 g Push/Pull Force Gauge 40 X .500LBS.Capacity
₹122,000.00INR / Each
Wagner Instruments FDK-32 Push/Pull Force Gauge 32 X .250 Oz.Capacity 9394962 Mfr # FDK-32
COMPARE
0.25 oz / 10 g Push/Pull Force Gauge 32 X .250 Oz.Capacity
₹122,000.00INR / Each
Wagner Instruments FDK-160 Push/Pull Force Gauge 10LBS.X 2oz.Capacity 9394963 Mfr # FDK-160
COMPARE
2 oz / 50 g Push/Pull Force Gauge 10LBS.X 2oz.Capacity
₹122,000.00INR / Each
Wagner Instruments FDK-60 Force Gauge 60Lbs Push/Pull Force Gauge 60 X .500LBS.Capacity 9394964 Mfr # FDK-60
COMPARE
0.500 lb / 250 g Force Gauge 60Lbs Push/Pull Force Gauge 60 X .500LBS.Capacity
₹122,000.00INR / Each
Showing 1- 9 of 9