W.S. Tyler Ro-Tap Shakers for 8 inch Diameter Sieves
Reproduce the rotating and tapping motion of hand-sieving but with more uniform motion for highly reproducible results
  • Ro-tap® shakers for 8'' dia sieves hold up to six full-height or 13 half height sieves with top cover and receiving pan
Shakers are actuated by a built-in, 99-minute digital timer with 0.1 second accuracy. They are powered by a 1/4-hp motor and include 5-ft cord (110 VAC models also include U.S. three-prong plug).
1,022,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) RPM Sieves Size Accepted (OD)
Availability Pricing
W.S. Tyler Ro-Tap Shaker for 8" Diameter Sieves, 110 VAC, 60 Hz 5998660 Mfr # RX-29
COMPARE
110 1750 8"
₹1,022,000.00INR / Each
W.S. Tyler Ro-Tap Shaker for 8" Diameter Sieves, 220 VAC, 50 hz 5998662 Mfr # RX-29-10
COMPARE
220 1450 8
₹1,022,000.00INR / Each