W.S. Tyler Ro-Tap Shakers for 12" Diameter Sieves
Reproduce the rotating and tapping motions of hand-sieving but with more uniform motion for highly reproducible results
  • Ro-tap® shakers for 12'' dia sieves hold up to four full-height or ten half height sieves with top cover and receiving pan
Shakers are actuated by a built-in, 99-minute digital timer with 0.1 second accuracy. They are powered by a 1/4-hp motor and include 5-ft cord (110 VAC models also include U.S. three-prong plug).
1,364,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) RPM Sieves Size Accepted (OD)
Availability Pricing
W.S. Tyler Ro-Tap Shaker for 12" Diameter Sieves, 110 VAC, 60 Hz 5999660 Mfr # RX-30
COMPARE
110 1750 12"
₹1,364,000.00INR / Each
W.S. Tyler Ro-Tap Shaker for 12" Diameter Sieves, 220 VAC, 50 Hz 5999662 Mfr # RX-30-10
COMPARE
220 1450 12"
₹1,364,000.00INR / Each