Vivaspin 6 Centrifugal Concentrators with PES MWCO Membranes
Broad range of process alternatives for the filtration and concentration of biological samples
  • Disposable ultrafiltration devices concentrate and purify biological samples to concentrations over 100x in a single spin!
  • Concentrators feature twin vertical membranes—increased surface area provides faster filtration
  • Polyethersulfone membrane is excellent for most solutions when recovery is most important
The Vivaspin 6 features twin vertical membranes for unparalleled filtration speeds and 100x plus concentrations. Remaining volume is easy to read off the printed scale on the side of the concentrator and the modified dead stop pocket further simplifies direct pipette recovery of the final concentrate.
43,200.00INR / Pkg of 25
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Molecular Weight Cut Off (Mwco) (Dalton) Description
Availability Pricing
Sartorius Vivaspin 6 Centrifugal Concentrators, PES Membrane 5000 Da; 25/Pk 3622816 Mfr # VS0611
COMPARE
5000 Centrifugal Concentrators, PES Membrane 5000 Da; 25/Pk
₹43,200.00INR / Pkg of 25
Sartorius Vivaspin 6 Centrifugal Concentrators, PES Membrane 30,000 Da; 25/Pk 3622820 Mfr # VS0621
COMPARE
30000 Centrifugal Concentrators, PES Membrane 30,000 Da; 25/Pk
₹43,200.00INR / Pkg of 25
Sartorius Vivaspin 6 Centrifugal Concentrators, PES Membrane 50,000 Da; 25/Pk 3622822 Mfr # VS0631
COMPARE
50000 Centrifugal Concentrators, PES Membrane 50,000 Da; 25/Pk
₹43,200.00INR / Pkg of 25
Sartorius Vivaspin 6 Centrifugal Concentrators, PES Membrane 100,000 Da; 25/Pk 3622824 Mfr # VS0641
COMPARE
100000 Centrifugal Concentrators, PES Membrane 100,000 Da; 25/Pk
₹43,200.00INR / Pkg of 25
Sartorius Vivaspin 6 Centrifugal Concentrators, PES Membrane 300,000 Da; 25/Pk 3622826 Mfr # VS0651
COMPARE
300000 Centrifugal Concentrators, PES Membrane 300,000 Da; 25/Pk
₹43,200.00INR / Pkg of 25
Sartorius Vivaspin 6 Centrifugal Concentrators, PES Membrane 1,000,000 Da; 25/Pk 3622828 Mfr # VS0661
COMPARE
1000000 Centrifugal Concentrators, PES Membrane 1,000,000 Da; 25/Pk
₹43,200.00INR / Pkg of 25