Vivaspin 15 R Centrifugal Concentrators with PES MWCO Membranes
Broad range of process alternatives for the filtration and concentration of biological samples
  • Disposable ultrafiltration devices concentrate and purify biological samples to concentrations over 100x in a single spin!
  • Concentrators feature twin vertical membranes—increased surface area provides faster filtration
  • Polyethersulfone membrane is excellent for most solutions when recovery is most important
This product has a modified regenerated cellulose membrane specially designed for ''sticky'' samples. This membrane has ultimate recovery at low adsorption (95 to 98%) and very small hold up volume (<20 µl). Ideal for desalting and concentration of Ig fractions.
37,900.00INR / Pkg of 12
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Molecular Weight Cut Off (Mwco) (Dalton)
Availability Pricing
Sartorius Vivaspin 15 R Centrifugal Concentrators, PES Membrane 5,000 Da; 12/Pk 3622832 Mfr # VS15RH11
COMPARE
5000
₹37,900.00INR / Pkg of 12
Sartorius Vivaspin 15 R Centrifugal Concentrators, PES Membrane 10,000 Da; 12/Pk 3622834 Mfr # VS15RH01
COMPARE
10000
₹37,900.00INR / Pkg of 12
Sartorius Vivaspin 15 R Centrifugal Concentrators, PES Membrane 30,000 Da; 12/Pk 3622836 Mfr # VS15RH21
COMPARE
30000
₹37,900.00INR / Pkg of 12