Vespel® Graphite Straight Ferrules
Ideal for GC/MS interface applications
  • Manufactured of 60% Vespel and 40% Graphite
Vespel®/Graphite Ferrules are one-piece design requiring no back ferrule. Maximum temperature: 400°C. Recommended for GC/MS interface applications. Reusable – does not seize into column.
6,400.00 - ₹7,400.00INR / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
VESPel Graphite Straight Ferrules for Packed Columns, 60% Vespel and 40% Graphite, 1/16"; 10/Pk WW-01993-50 Mfr # 213100
COMPARE
/Graphite Straight Ferrules for Packed Columns, 1/16\"
₹6,400.00INR / Pkg of 10
VESPel Graphite Straight Ferrules for Packed Columns, 60% Vespel and 40% Graphite, 1/8"; 10/Pk WW-01993-51 Mfr # 213200
COMPARE
/Graphite Straight Ferrules for Packed Columns, 1/8\"
₹7,200.00INR / Pkg of 10
VESPel Graphite Straight Ferrules for Packed Columns, 60% Vespel and 40% Graphite, 1/4"; 10/Pk WW-01993-52 Mfr # 213400
COMPARE
/Graphite Straight Ferrules for Packed Columns, 1/4\"
₹7,400.00INR / Pkg of 10