Versilon® PEEK Capillary Tubing

High-performance PEEK tubing for chemically resistant, high-pressure fluid handling
  • Excellent chemical and corrosion resistance provides a reliable alternative to stainless steel in demanding applications
  • Engineered to withstand high pressures and elevated temperatures for HPLC and critical fluid handling
  • Ultra-low extractables help maintain sample integrity in sensitive analytical workflows
  • Durable, dimensionally stable construction ensures consistent performance under continuous use
Versilon® PEEK tubing is designed for high-performance applications where chemical resistance, purity, and pressure stability are critical. Engineered to meet the demands of high-performance liquid chromatography (HPLC) and similar workflows, this tubing offers superior corrosion resistance compared to stainless steel while maintaining the structural integrity needed for high-pressure operation. Its ultra-low extractables and resistance to steam exposure make it well-suited for sensitive analytical and laboratory environments. With excellent mechanical strength, dimensional stability, and continuous service capability at elevated temperatures, Versilon® PEEK tubing provides a dependable solution for consistent, long-term performance in demanding fluid handling systems.
312,000.00 - ₹790,000.00INR / Pkg of 1

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (mm) Tubing OD (mm) Wall Thickness (mm)
Availability Pricing
Versilon PEEK®  Capillary Tubing, 1/50" ID x 1/16" OD, 250 ft 4852601 Mfr # TSPK38-0062-021
COMPARE
0.5 1.6 0.5
₹317,000.00INR / Pkg of 1
Versilon PEEK®  Capillary Tubing, 1/32" ID x 1/16" OD, 250 ft 4852602 Mfr # TSPK38-0062-016
COMPARE
0.8 1.6 0.4
₹312,000.00INR / Pkg of 1
Versilon PEEK®  Capillary Tubing, 1/16" ID x 1/8" OD, 250 ft 4852603 Mfr # TSPK38-0125-031
COMPARE
1.6 3.2 0.8
₹790,000.00INR / Pkg of 1
Versilon PEEK®  Capillary Tubing, 5/64" ID x 1/8" OD, 250 ft 4852604 Mfr # TSPK38-0125-023
COMPARE
2 3.2 0.6
₹790,000.00INR / Pkg of 1
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat