Versilon® Flexible PVDF Tubing
Flexible PVDF tubing for high-purity, chemically resistant fluid handling
  • Combines fluoropolymer chemical resistance with added flexibility for easier routing and installation
  • High purity and low extractables support sensitive and regulated applications
  • Strong, abrasion-resistant construction provides durability in demanding environments
  • Resistant to UV, radiation, and harsh chemicals for long-term performance
Versilon® PVDF tubing is designed for applications requiring a balance of chemical resistance, flexibility, and mechanical durability. Made from a PVDF copolymer, it delivers the corrosion resistance expected from fluoropolymer materials while offering greater flexibility than standard PVDF tubing, making it easier to install and route in complex systems. Its high purity and low extractables make it well-suited for sensitive applications in industries such as semiconductor processing, water purification, and chemical handling. With strong resistance to abrasion, UV exposure, and aggressive media, Versilon® PVDF tubing provides a reliable solution for long-term performance in demanding environments.
21,800.00 - ₹222,000.00INR / Pkg of 1
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Versilon Flexible PVDF Tubing, 1/16" ID x 1/8" OD, 50 ft 4852639 Mfr # TSPVF8-0125-031-50
COMPARE
1/16 1/8 1/32
₹21,800.00INR / Pkg of 1
Versilon Flexible PVDF Tubing, 1/8" ID x 3/16" OD, 50 ft 4852640 Mfr # TSPVF8-0187-031-50
COMPARE
1/8 3/16 1/32
₹23,300.00INR / Pkg of 1
Versilon Flexible PVDF Tubing, 11/64" ID x 1/4" OD, 50 ft 4852641 Mfr # TSPVF8-0250-040-50
COMPARE
11/64 1/4 1/25
₹47,500.00INR / Pkg of 1
Versilon Flexible PVDF Tubing, 1/8" ID x 1/4" OD, 50 ft 4852642 Mfr # TSPVF8-0250-062-50
COMPARE
1/8 1/4 1/16
₹52,200.00INR / Pkg of 1
Versilon Flexible PVDF Tubing, 3/16" ID x 5/16" OD, 50 ft 4852643 Mfr # TSPVF8-0312-062-50
COMPARE
3/16 5/16 1/16
₹72,400.00INR / Pkg of 1
Versilon Flexible PVDF Tubing, 5/16" ID x 3/8" OD, 50 ft 4852644 Mfr # TSPVF8-0375-031-50
COMPARE
5/16 3/8 1/32
₹91,400.00INR / Pkg of 1
Versilon Flexible PVDF Tubing, 3/8" ID x 1/2" OD, 50 ft 4852645 Mfr # TSPVF8-0500-062-50
COMPARE
3/8 1/2 1/16
₹125,000.00INR / Pkg of 1
Versilon Flexible PVDF Tubing, 1/2" ID x 5/8" OD, 50 ft 4852646 Mfr # TSPVF8-0625-062-50
COMPARE
1/2 5/8 1/16
₹157,000.00INR / Pkg of 1
Versilon Flexible PVDF Tubing, 5/8" ID x 3/4" OD, 50 ft 4852647 Mfr # TSPVF8-0750-062-50
COMPARE
5/8 3/4 1/16
₹195,000.00INR / Pkg of 1
Versilon Flexible PVDF Tubing, 3/4" ID x 7/8" OD, 50 ft 4852648 Mfr # TSPVF8-0875-062-50
COMPARE
3/4 7/8 1/16
₹222,000.00INR / Pkg of 1
Showing 1- 10 of 10