Versilon® 2001 Plasticizer-Free Chemical-Resistant Tubing
Superior chemical resistance to a wide range of aggressive fluid—even use with polar solvents
  • Plasticizer-free and oil-free tubing provides contamination-free fluid transfer
  • Long service life in peristaltic pumps
  • Clear wall provides visual monitoring of flow
  • Meets FDA 21 CFR 177.2600 requirements for food contact
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Use the Versilon® chemical-resistant tubing with a wide range of aggressive chemicals, including polar solvents. Durable Versilon 2001 tubing provides a long service life and does not degrade over time, leading to less downtime and tubing changes from tubing failure.
Great flexibility, excellent chemical resistance, and long service life make this tubing ideal for peristaltic pump applications. Plasticizer-free formulation does not contaminate fluids and will not yield any taste, making it ideal for food and beverage applications and water purification line. Clear tubing allows easy monitoring of the flow, so you can control or adjust the flow during the process.
11,700.00 - ₹138,000.00INR / Pkg of 1
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Versilon 2001 Plasticizer-Free Chemical-Resistant Tubing, 1/16" ID x 3/16" OD; 50 Ft 5012251 Mfr # AE300003
COMPARE
1/16 3/16 1/16
₹11,700.00INR / Pkg of 1
Versilon 2001 Plasticizer-Free Chemical-Resistant Tubing, 1/8" ID x 1/4" OD; 50 Ft 5012252 Mfr # AE300007
COMPARE
1/8 1/4 1/16
₹18,500.00INR / Pkg of 1
Versilon 2001 Plasticizer-Free Chemical-Resistant Tubing, 3/16" ID x 5/16" OD; 50 Ft 5012253 Mfr # AE300012
COMPARE
3/16 5/16 1/16
₹20,700.00INR / Pkg of 1
Versilon 2001 Plasticizer-Free Chemical-Resistant Tubing, 1/4" ID x 3/8" OD; 50 Ft 5012254 Mfr # AE300017
COMPARE
1/4 3/8 1/16
₹22,800.00INR / Pkg of 1
Versilon 2001 Plasticizer-Free Chemical-Resistant Tubing, 5/16" ID x 7/16" OD; 50 Ft 5012255 Mfr # AE300022
COMPARE
5/16 7/16 1/16
₹25,000.00INR / Pkg of 1
Versilon 2001 Plasticizer-Free Chemical-Resistant Tubing, 3/8" ID x 1/2" OD; 50 Ft 5012256 Mfr # AE300027
COMPARE
3/8 1/2 1/16
₹27,800.00INR / Pkg of 1
Versilon 2001 Plasticizer-Free Chemical-Resistant Tubing, 1/2" ID x 3/4" OD; 50 Ft 5012257 Mfr # AE300038
COMPARE
1/2 3/4 1/8
₹62,700.00INR / Pkg of 1
Versilon 2001 Plasticizer-Free Chemical-Resistant Tubing, 5/8" ID x 7/8" OD; 50 Ft 5012258 Mfr # AE300046
COMPARE
5/8 7/8 1/8
₹73,500.00INR / Pkg of 1
Versilon 2001 Plasticizer-Free Chemical-Resistant Tubing, 3/4" ID x 1" OD; 50 Ft 5012259 Mfr # AE300053
COMPARE
3/4 1 1/8
₹79,200.00INR / Pkg of 1
Versilon 2001 Plasticizer-Free Chemical-Resistant Tubing, 1" ID x 1-3/8" OD; 50 Ft 5012260 Mfr # AE300064
COMPARE
1 1 3/8 3/16
₹138,000.00INR / Pkg of 1
Showing 1- 10 of 10