Velp® Multiposition Digital Stirrers
Stays cool, even after days of continuous use
  • SpeedServo™ technology ensures constant speed even when the viscosity changes
  • Soft start reduces splashing
Excellent speed control and synchronized stirring ensure that the same speed is achieved on each position even at low speeds. The low-profile, lightweight design makes these stirrers easy to move while saving valuable bench space. Stainless steel top plate construction is easy to clean and provides high chemical resistance. Featuring adjustable time controlled reverse operation, timer, and a bright digital display for accurate speed monitoring, these stirrers will provide years of worry-free operation.
247,460.00 - ₹361,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
No. Of Stirring Positions Power (VAC)
Availability Pricing
Velp F203A0179 Multiposition Digital Stirrer, 6-position, 120-240V 8657957 Mfr # F203A0179
COMPARE
6 110 to 240
₹361,000.00INR / Each
Velp F203A0180 Multiposition Digital Stirrer, 15-position, 120-240V 8657958 Mfr # F203A0180
COMPARE
15 110 to 240
₹247,460.00INR / Each