Velp Laboratory Mixers

Chemical-resistant sealed housing withstands aggressive liquids and vapors
  • Gentle start-up through optimal progression of stirring speed
  • Keyless chuck accommodates shafts with up to 3/8” (10 mm) diameter
  • Advanced safety features include overload, overcurrent, and overtemperature protection
These versatile and lightweight mixers feature a unique technopolymer structure that provides chemical resistance and rust protection. The user-friendly design and slim casting save you valuable laboratory bench space. The sealed housing shields stirrer from aggressive liquids and offers premium resistance from acids, bases, and solvents.

Self-locking keyless chuck and a through-shaft design let you easily adjust impeller depth to fit your vessel without repositioning the mixer. Robust motor permits continuous operation with protection
from overload, overcurrent, and overtemperature. Designed for a long and reliable service life.
97,300.00 - ₹190,000.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Max Viscosity (cps) Max Volume (Liters)
Availability Pricing
Velp F201A0152 Laboratory Mixer, 1000 cps, 50-1300 rpm; 80-260 VAC, 50/60 Hz WW-50200-00 Mfr # F201A0152
COMPARE
1300 1000 15
₹97,300.00INR / Each
Velp F201A0151 Laboratory Mixer, 25,000 cps WW-50200-02 Mfr # F201A0151
COMPARE
2000 25000 25
₹131,000.00INR / Each
Velp F201A0156 Laboratory Mixer, 50,000 cps WW-50200-04 Mfr # F201A0156
COMPARE
2000 50000 40
₹166,000.00INR / Each
Velp F201A0150 Laboratory Mixer, 100,000 cps WW-50200-06 Mfr # F201A0150
COMPARE
1200 100000 70
₹190,000.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat