Velp Kjeldahl Systems

High degree of precision, reproducibility, and versatility
  • Configure a complete system for Kjeldahl analysis by combining a digester with distillation/titration units
  • Suitable for food and beverage, environmental, chemical, and pharmaceutical applications
  • Produce results according to standards such as AOAC, EPA, DIN, and ISO while ensuring conformity to GLP
DKL 8 Fully Automated Digestion Unit

Operational right out of the box!
35% reduction in power consumption, cutting CO2 emissions
Rapid heatup time—only 22 minutes to reach 420°C
Slim space-saving design saves valuable bench space

The DKL 8 unit provides simultaneous digestion of eight 250-mL samples to prepare for the distillation phase. Sample protection is tightly controlled with temperature precision and stability of ±0.5°C. Full automation simplifies operation and improves the reproducibility of your results.

After completing setup, just press the "start" button and walk away! All regular functions are fully automated, including lowering the test tubes into the heating block, monitoring the operation, storing run data, lifting the tubes to expedite cooling and separating the suction manifold from the tube rack, enabling the rack to be slid out and replaced by another to start the next run. No need to handle the tubes when hot, increasing user safety.

A 54-program library lets you select your application and start work immediately. Simple 6-button keypad, clear graphic display, and multiple languages quickly guide you in planning and monitoring the digestion phases. Plus, the USB interface offers a convenient method to download data to your PC.
Order the scrubber and vacuum pump to neutralize fumes produced during digestion for operator and environmental safety.

What’s included: lift, drip tray, suction cap, stainless steel sample rack with cover, USB cable, and eight 250-mL digestion tubes.

UDK Series Distillation Units

Fast and frequent analyses with no heating delays between runs

Compact units offer different levels of automation, from simple distillation, to distillation with automatic reagents addition and residues aspiration, up to fully automated system with integrated colorimetric titration. Ideal for applications such as determining ammoniacal nitrogen, protein nitrogen (Kjeldahl or direct alkaline distillation), nitric nitrogen (after reduction), phenols, volatile fatty acids, cyanides, alcohol content and Devarda nitrogen determination.

Unique steam generator combined with a technopolymer splash head for performing efficient and reliable steam distillations. Designed with a strong and chemical-resistant structure, these units are designed to perform reliable analysis for many years. Innovative features like safety lever, protective door, automatic electric shut down, water flow rate detector and test tube sensors improve the safety level.
What''s included: test tube, pincer for tube, collecting flask, and inlet and discharge tubes.
186,000.00 - ₹983,000.00INR / Each

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Condenser Type Evaporation Flask Size Max Speed (rpm)
Availability Pricing
Velp DLK-8 Automatic Digestion Unit 1595620 Mfr # S30100200
COMPARE
Call For Price
Velp JP PUMP JP Recirculating Water Pump for DLK-8 1595621 Mfr # F30620198
COMPARE
₹305,000.00INR / Each
Velp SMS SCRUBBER SMS Scrubber for DLK-8 1595622 Mfr # F307C0199
COMPARE
₹186,000.00INR / Each
Velp UDK 139 Distillation Unit 1595624 Mfr # F30200130
COMPARE
₹983,000.00INR / Each
Velp F30200140 UDK149 Distillation Unit 230V/50-60Hz 1595659 Mfr # F30200140
COMPARE
Call For Price
Velp UDK169 Distillation unit 230V/50-60Hz 1595660 Mfr # S30200160 (UDK169 + AUTOSAMPLER)
COMPARE
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat