Velp AREX 5 Digital Stirring Hot Plates
Ensures precise thermoregulation of a sample
  • Bright, easy-to-read digital display
  • Message displays when plate get hot
  • Can reach maximum temperature of 310 °C (590 °F)
  • Stirring speed up to 1500 rpm
  • Able to stir up to 20 liters
  • IP42 certified
  • 2 year warranty
The AREX 5 digital stirring hot plates are designed for temperature-sensitive laboratory applications where precise sample thermoregulation is required for maximum reproducibility. The units are safe to use. A “hot” message is displayed on the unit when the temperature of the plate goes above 50 °C (122 °F) and heating function is not active. Internal electronic parts are protected by the insulated disk and die-cast aluminum housing.
The easy-to-read digital display allows you to clearly read the values shown while monitoring the temperature and stirring speed.
The 135 mm diameter CerAlTop™ aluminum alloy heating top plate ensures temperature homogeneity and optimum heat distribution across the entire surface. The plate is resistant to thermal stress and shock. A special ceramic coating ensures high resistance to chemical agents and surface abrasion. The magnetic coupling supports reactions with multiple vials ensuring constant speed in all positions.
The powerful brushless motor can stir up to 20 liters from 50 to 1500 rpm. SpeedServo™ torque compensation technology ensures you constant speed as viscosity changes.
Some models are available with an external PT100 temperature probe or VTF temperature controller which can heat up the medium up to 300 °C (572 °F) with an accuracy of ± 0.5 °C. Choose the model that is right for you.
222,000.00 - ₹342,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Stirring Volume (Liters) Max Temperature (° C) Power (VAC)
Availability Pricing
Velp AREX 5  Digital Stirring Hot Plate; 115V 8657892 Mfr # F20510580
COMPARE
20 310 115
₹222,000.00INR / Each
Velp AREX 5  Digital Stirring Hot Plate; 230V 8657893 Mfr # F20500580
COMPARE
20 310 230
₹222,000.00INR / Each
Velp AREX 5  Digital Stirring Hot Plate with PT100 Temperature Probe; 115V 8657894 Mfr # SA20510580
COMPARE
20 310 115
₹243,000.00INR / Each
Velp AREX 5  Digital Stirring Hot Plate with PT100 Temperature Probe; 230V 8657895 Mfr # SA20500580
COMPARE
20 310 230
₹243,000.00INR / Each
Velp AREX 5  Digital Stirring Hot Plate with PT100 Temperature Probe, Rod and Clamp; 115V 8657896 Mfr # SC20510580
COMPARE
20 310 115
₹258,000.00INR / Each
Velp AREX 5  Digital Stirring Hot Plate with PT100 Temperature Probe, Rod and Clamp; 230V 8657897 Mfr # SC20500580
COMPARE
20 310 230
₹258,000.00INR / Each
Velp AREX 5  Digital Stirring Hot Plate with VTF Temperature Probe; 115V 8657898 Mfr # SB20510580
COMPARE
20 300 115
₹342,000.00INR / Each
Velp AREX 5  Digital Stirring Hot Plate with VTF Temperature Probe; 230V 8657899 Mfr # SB20500580
COMPARE
20 300 230
₹342,000.00INR / Each
Showing 1- 8 of 8