Value Plastics Male Luer Integral Lock Ring to UNF Thread Adapters
Prevent leaks with integral lock ring mechanism
  • Easily transition from a luer to thread fitting
  • Luer fittings allow for a simple yet secure twist-lock connection
  • UNF and specialty thread fittings are designed to create a tight seal under pressure
Value Plastics Male Luer Integral Lock Ring to UNF Thread Adapters are designed to easily transition your fluid transfer system from a luer to thread connection. Luer fittings ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The integral lock ring is a feature built into the male luer fitting that ensures the fittings remain firmly connected until intentionally disconnected. UNF and specialty thread fittings are finer, having more threads per inch, and are commonly used in automotive, aerospace, and hydraulic systems applications. Threaded fittings are designed to create a tight seal when assembled, preventing leakage of fluids or gases under pressure. These adapters are available in various materials to accommodate various applications and compliance requirements.
3,800.00 - ₹5,000.00INR / Pkg of 25
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Fitting Material
Availability Pricing
Value Plastics Luer to Threaded Fitting, Straight Adapter, White Nylon, Male Luer Integral Lock Ring x 1/4-28 UNF (M) Thread; 25/Pk 5012087 Mfr # SMTLL-1
COMPARE
Nylon
₹3,800.00INR / Pkg of 25
Value Plastics Luer to Threaded Fitting, Straight Adapter, Polypropylene, Male Luer Integral Lock Ring x 10-32 (M) Special Tapered Thread; 25/Pk 5012089 Mfr # XMTLL-6005
COMPARE
Polypropylene
₹5,000.00INR / Pkg of 25