Value Plastics Female Luer to 200 Series Barb
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
  • Rotating locks minimize twisting and kinking of tubing during positioning
  • Tapered cone seals are reliable and uncomplicated
  • Facilitate single-use configurations that minimize downtime
These luer fittings come in many configurations and several materials, joining tubing from 1/16” (1.6 mm) ID to 1/4” (6.4 mm) ID. The taper of the female luer is designed to match the taper of the male luer, creating a secure and leak-free connection when the two parts are joined together. The 200 series barb offers outstanding performance characteristics when used with flexible tubing. Tubing is expanded by 50% above its nominal inside diameter, permitting high-pressure capabilities. Because the fittings are made to ISO standards that are well understood globally, it ensures proper interconnections and often removes the need to retrofit*.
Options include nylon, polycarbonate, or polypropylene resin.
Common features of nylon include a high mechanical strength, great balance of stiffness and toughness, good performance at high temperatures, good electrical and flammability properties, and resistance against abrasion or chemicals. It can be sterilized using gamma radiation, Ethylene oxide (EtO), or autoclave.
Polypropylene is clarified and produced without any animal derived components or phthalates. It can be sterilized using gamma radiation, Ethylene oxide (EtO), E-beam, or autoclave.
Polycarbonate is transparent and features high impact, chemical, UV, and electrical resistance. Sterilization is only recommended via ethylene oxide (EtO). Gamma radiation up to 10 Mrads is possible with minor loss of physical properties – it will discolor to a yellow-green hue. Autoclave is not recommended and may crack the fitting.
2,400.00 - ₹3,500.00INR / Pkg of 25
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Female Luer Thread x 200 Series Barb 1/16" ID; 25/Pk 5012037 Mfr # FTLL210-6005
COMPARE
1/16 Polypropylene
₹3,100.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Female Luer Thread x 200 Series Barb 1/8" ID; 25/Pk 5012038 Mfr # FTLL230-1
COMPARE
1/8 Nylon
₹2,500.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Female Luer Thread x 200 Series Barb 3/16" ID; 25/Pk 5012039 Mfr # FTLL250-1
COMPARE
3/16 Nylon
₹2,700.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Female Luer Thread x 200 Series Barb 3/32" ID; 25/Pk 5012044 Mfr # FTLL220-6005
COMPARE
3/32 Polypropylene
₹3,100.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Female Luer Thread x 200 Series Barb 5/32" ID; 25/Pk 5012045 Mfr # FTLL240-6005
COMPARE
5/32 Polypropylene
₹3,500.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Female Luer Thread x 200 Series Barb 1/16" ID; 25/Pk 5012049 Mfr # FTLL210-1
COMPARE
1/16 Nylon
₹2,400.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Clear Polycarbonate, Female Luer Thread x 200 Series Barb 3/32" ID; 25/Pk 5012050 Mfr # FTLL220-9
COMPARE
3/32 Polycarbonate
₹2,900.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Clear Polycarbonate, Female Luer Thread x 200 Series Barb 5/32" ID; 25/Pk 5012051 Mfr # FTLL240-9
COMPARE
5/32 Polycarbonate
₹3,200.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Female Luer Thread x 200 Series Barb 1/8" ID; 25/Pk 5012055 Mfr # FTLL230-6005
COMPARE
1/8 Polypropylene
₹3,200.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Clear Polycarbonate, Female Luer Thread x 200 Series Barb 1/8" ID; 25/Pk 5012059 Mfr # FTLL230-9
COMPARE
1/8 Polycarbonate
₹2,900.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Female Luer Thread x 200 Series Barb 3/32" ID; 25/Pk 5012063 Mfr # FTLL220-1
COMPARE
3/32 Nylon
₹2,400.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Female Luer Thread x 200 Series Barb 5/32" ID; 25/Pk 5012064 Mfr # FTLL240-1
COMPARE
5/32 Nylon
₹2,700.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Clear Polycarbonate, Female Luer Thread x 200 Series Barb 1/16" ID; 25/Pk 5012067 Mfr # FTLL210-9
COMPARE
1/16 Polycarbonate
₹2,900.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Clear Polycarbonate, Female Luer Thread x 200 Series Barb 3/16" ID; 25/Pk 5012068 Mfr # FTLL250-9
COMPARE
3/16 Polycarbonate
₹3,200.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Female Luer Thread x 200 Series Barb 3/16" ID; 25/Pk 5012070 Mfr # FTLL250-6005
COMPARE
3/16 Polypropylene
₹3,400.00INR / Pkg of 25
Showing 1- 15 of 15